476447
1-Iodo-3-methylbutane
97%
Synonym(s):
Isoamyl iodide, Isopentyl iodide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH2CH2I
CAS Number:
Molecular Weight:
198.05
Beilstein:
1730990
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.492 (lit.)
bp
149 °C (lit.)
density
1.497 g/mL at 25 °C (lit.)
functional group
alkyl halide
iodo
SMILES string
CC(C)CCI
InChI
1S/C5H11I/c1-5(2)3-4-6/h5H,3-4H2,1-2H3
InChI key
BUZZUHJODKQYTF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F
Flash Point(C)
40 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Vibrational analysis of alkyl iodides: 1-iodo-2-methylpropane and 1-iodo-3-methylbutane.
Crowder GA and Jalilian MR .
Journal of Molecular Structure, 49(2), 287-292 (1978)
Ketoreductases in the synthesis of valuable chiral intermediates: application in the synthesis of α-hydroxy β-amino and β-hydroxy γ-amino acids.
Kambourakis S and Rozzell JD.
Tetrahedron, 60(3), 663-669 (2004)
The far ultra-violet spectra of some branched chain iodo-alkanes, iodo-cyclo-alkanes, fluoro-iodo-alkanes and iodo-alkenes.
Boschi RA and Salahub DR.
Molecular Physics, 24(4), 735-752 (1972)
Ultrasound mediated synthesis of a few naturally occurring compounds.
Singh J, et al.
Indian J. Chem. B, 40(5), 386-390 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service