Skip to Content
Merck
All Photos(1)

Key Documents

473111

Sigma-Aldrich

Lithium-6Li2 carbonate

95 atom % 6Li

Synonym(s):

Carbolithium-6, Carbonic acid lithium-6 salt, Lithium carbonate-6Li2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
6Li2CO3
CAS Number:
Molecular Weight:
72.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

95 atom % 6Li

Quality Level

form

solid

mp

720 °C (lit.)

mass shift

M-2

SMILES string

[6Li+].[6Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2/i;2*1-1

InChI key

XGZVUEUWXADBQD-OUDCDBHYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service