Skip to Content
Merck
All Photos(2)

Documents

471674

Sigma-Aldrich

(1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

99%

Synonym(s):

L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH(OH)CH(NH2)CH2OH
CAS Number:
Molecular Weight:
212.20
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]23/D +31°, c = 1 in 6 M HCl

mp

163-166 °C (lit.)

SMILES string

N[C@@H](CO)[C@@H](O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C9H12N2O4/c10-8(5-12)9(13)6-1-3-7(4-2-6)11(14)15/h1-4,8-9,12-13H,5,10H2/t8-,9-/m0/s1

InChI key

OCYJXSUPZMNXEN-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induced synthesis of phosphatases in Anacystis nidulans by p-NO2-phenylserinal.
M L Muñoz-Calvo et al.
The Journal of antibiotics, 33(5), 520-522 (1980-05-01)
D K Zheng et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 25(6), 430-434 (1990-01-01)
Threo (-) or (+)-1-(p-nitrophenyl)-1,3-dihydroxypropylamine-2 was found to be a useful resolving agent for racemic gossypol. The optical and chemical stability of the condensate of enantiomeric gossypols with the titled amine and the great difference in the Rf values on TLC
D A Pearce et al.
Molecular genetics and metabolism, 66(4), 314-319 (1999-04-07)
The CLN3 gene, which encodes the protein whose absence is responsible for Batten disease, the most common inherited neurovisceral storage disease of childhood, was identified in 1995. The function of the protein, Cln3p, still remains elusive. We previously cloned the
W A Korfmacher et al.
Journal of chromatographic science, 28(5), 236-238 (1990-05-01)
The thermospray (TS) liquid chromatographic-mass spectrometric analysis of the antibiotic chloramphenicol and three related compounds is presented. The three additional compounds are dehydrochloramphenicol, aminodehydrochloramphenicol, and nitrophenylaminopropanediol. Baseline separation of the four compounds is achieved. The TS mass spectrum of each
High-performance liquid chromatographic determination of chloramphenicol and 2-amino-1-(p-nitrophenyl)-1,3-propanediol in pharmaceutical formulations.
M J LeBelle et al.
Journal of chromatography, 170(1), 282-287 (1979-02-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service