Skip to Content
Merck
All Photos(1)

Key Documents

467995

Sigma-Aldrich

Thiazolidine-2-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2S
CAS Number:
Molecular Weight:
133.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

176 °C (lit.)

SMILES string

OC(=O)C1NCCS1

InChI

1S/C4H7NO2S/c6-4(7)3-5-1-2-8-3/h3,5H,1-2H2,(H,6,7)

InChI key

ULSZVNJBVJWEJE-UHFFFAOYSA-N

General description

Thiazolidine-2-carboxylic acid (β-thiaproline) is a proline analog. It is an important building block of β-lactam antibiotics. Its X-ray photoelectron spectra has been investigated. It has been reported as a physiological substrate of hog kidney D-amino acid oxidase. Thiazolidine-2-carboxylic acid can be synthesized from cysteamine and glyoxylate.

Application

Thiazolidine-2-carboxylic acid may be used in the synthesis of azabicycloadducts and 5-aryl-2,3-dihydropyrrolo[2,1-b]thiazole-6,7-dimethanol 6,7-bis(isopropylcarbamates).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Busiello et al.
Biochimica et biophysica acta, 564(2), 311-321 (1979-09-27)
Thiazolidine-2-carboxylic acid, or beta-thiaproline, is a proline analog in which the beta methylene group of proline is substituted by a sulfur atom. It has been deomonstrated that beta-thiaproline is activated and transferred to tRNAPro by Escherichia coli and rat liver
L Włodek et al.
Biochemical pharmacology, 46(1), 190-193 (1993-07-06)
2-Substituted thiazolidine-4(R)-carboxylic acids (TD) were found to increase the concentration of non-protein sulphydryls (NPSH) and the activity of rhodanese (thiosulphate sulphurtransferase, EC 2.8.1.1) and 3-mercaptopyruvate sulphurtransferase (EC 2.8.1.2) in mouse liver. These properties suggest TDs are potentially hepatoprotective compounds. However
Synthesis and chromatographic properties of selenazolidine-2-carboxylic acid (beta-selenaproline).
C De Marco et al.
The Italian journal of biochemistry, 28(2), 104-110 (1979-03-01)
C L Burns et al.
Biochemical and biophysical research communications, 125(3), 1039-1045 (1984-12-28)
Adducts of glyoxylate with L-cysteine or L-cysteinylglycine were found to be excellent substrates at low concentrations for beef kidney D-aspartate oxidase. Evidence is presented that cis-thiazolidine-2,4-dicarboxylate and its glycine amide are the actual substrates, and that both are converted in
P F Fitzpatrick et al.
The Journal of biological chemistry, 257(3), 1166-1171 (1982-02-10)
A mixture of cysteamine and glyoxylate, proposed by Hamilton et al. to form the physiological substrate of hog kidney D-amino acid oxidase (Hamilton, G. A., Buckthal, D. J., Mortensen, R. M., and Zerby, K. W. (1979) Proc. Natl. Acad. Sci.

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service