Skip to Content
Merck
All Photos(2)

Key Documents

466212

Sigma-Aldrich

1,1,2-Trichloroethane

contains ≤3% 2-propanol as stabilizer, 97%

Synonym(s):

β-Trichloroethane, Vinyltrichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHCl2
CAS Number:
Molecular Weight:
133.40
Beilstein:
1731726
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

contains

≤3% 2-propanol as stabilizer

refractive index

n20/D 1.471 (lit.)

bp

110-115 °C (lit.)

density

1.435 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCC(Cl)Cl

InChI

1S/C2H3Cl3/c3-1-2(4)5/h2H,1H2

InChI key

UBOXGVDOUJQMTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,2-Trichloroethane (1,1,2-TCA), a chlorinated aliphatic hydrocarbon, is a common groundwater pollutant. Microbial detoxification of 1,1,2-TCA has been studied. Ni supported on mesoporous silica (SBA-16) catalyzed hydrodechlorination of 1,1,2-trichloroethane has been reported. Rotational isomers of 1,1,2-trichloroethane in the gaseous and liquid states have been investigated by the infrared absorption spectra.

Application

  • Advanced Material Development: Research into the synthesis of UiO-66 3D photonic crystals highlights the use of 1,1,2-Trichloroethane in the creation of new materials for efficient vapor detection, supporting advancements in sensor technology (Wang et al., 2022).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Infrared Absorption Investigation on the Rotational Isomerism of 1, 1, 2-Trichloroethane.
Kuratani K and Mizushima S-I.
J. Chem. Phys. , 22(8), 1403-1405 (1954)
Single-step preparation of Ni catalysts supported on mesoporous silicas (SBA-15 and SBA-16) and the effect of pore structure on the selective hydrodechlorination of 1, 1, 2-trichloroethane to VCM.
Park Y, et al.
Catalysis Today, 97(2), 195-203 (2004)
Siyan Zhao et al.
PloS one, 10(3), e0119507-e0119507 (2015-04-04)
1,1,2-trichloroethane (1,1,2-TCA) has become a common groundwater pollutant due to historically extensive utilization, improper disposal, as well as from incomplete dechlorination of 1,1,2,2-tetrachloroethane. Currently, limited information is available on microbial detoxification of 1,1,2-TCA. Desulfitobacterium sp. strain PR, which was isolated
Michio M Matsushita et al.
Journal of the American Chemical Society, 127(36), 12450-12451 (2005-09-08)
A bromide salt of a TTF-based cross-cyclophane-type donor displayed characteristic nonlinear voltage-biased conductivity. As the applied voltage was increased, the passing current was enhanced abruptly at 54 kV.cm-1, exhibiting hysteretic behavior. X-ray analysis of the current-induced low-resistance state of the
Ariel Grostern et al.
Applied and environmental microbiology, 72(1), 428-436 (2006-01-05)
Mixed anaerobic microbial subcultures enriched from a multilayered aquifer at a former chlorinated solvent disposal facility in West Louisiana were examined to determine the organism(s) involved in the dechlorination of the toxic compounds 1,2-dichloroethane (1,2-DCA) and 1,1,2-trichloroethane (1,1,2-TCA) to ethene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service