Skip to Content
Merck
All Photos(1)

Documents

461431

Sigma-Aldrich

(S)-(−)-N-(1-Phenylethyl)succinamic acid

98%

Synonym(s):

(S)-(−)-N-(α-Methylbenzyl)succinamidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCOCH2CH2CO2H
CAS Number:
Molecular Weight:
221.25
Beilstein:
3205511
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]24/D −111°, c = 2 in ethanol

mp

100-104 °C (lit.)

SMILES string

C[C@H](NC(=O)CCC(O)=O)c1ccccc1

InChI

1S/C12H15NO3/c1-9(10-5-3-2-4-6-10)13-11(14)7-8-12(15)16/h2-6,9H,7-8H2,1H3,(H,13,14)(H,15,16)/t9-/m0/s1

InChI key

WUEKFTPKHWMMIP-VIFPVBQESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service