Skip to Content
Merck
All Photos(2)

Key Documents

457701

Sigma-Aldrich

(S)-(−)-2-Methyl-CBS-oxazaborolidine solution

1 M in toluene

Synonym(s):

α,α-Diphenyl-L-prolinol methylboronic acid cycl-amide ester, (S)-1-Methyl-3,3-diphenyl-tetrahydro-pyrrolo[1,2c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methyltetrahydro-3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20BNO
CAS Number:
Molecular Weight:
277.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

concentration

1 M in toluene

bp

111 °C

density

0.929 g/mL at 25 °C

storage temp.

room temp

SMILES string

CB1OC([C@@H]2CCCN12)(c3ccccc3)c4ccccc4

InChI

1S/C18H20BNO/c1-19-20-14-8-13-17(20)18(21-19,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17H,8,13-14H2,1H3/t17-/m0/s1

InChI key

VMKAFJQFKBASMU-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-2-Methyl-CBS-oxazaborolidine solution [1M in toluene] may be used as a catalyst in the multi-step synthesis of (-)-diospongin A.
It may also be used in the preparation of:
  • (1S)-2-azido-1-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)ethanol
  • (R)-α-deuteriobenzyl alcohol
  • (R)-2-(1-hydroxyethyl)benzo[b]thiophene
Catalyst employed in an enantioselective synthesis of (S)-2-amino-1-phenylethanol.
Excellent catalyst for asymmetric reductions.
The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2-symmetrical ferrocenyl diols, and propargyl alcohols.

Other Notes

Precipitate may form in storage, but does not affect product quality. Gently heat product to 80-90 °C under an inert atmosphere to redissolve solids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organic Process Research & Development, 10, 893-893 (2006)
Enantioselective synthesis of (S)-salmeterol via asymmetric reduction of azidoketone by Pichia angusta.
Procopiou PA, et al.
Tetrahedron Asymmetry, 12(14), 2005-2008 (2001)
Diastereoselective photochromism of a bisbenzothienylethene governed by steric as well as electronic interactions.
Yokoyama Y, et al.
Journal of the American Chemical Society, 125(24), 7194-7195 (2003)
Stereoselective synthesis of (-)-diospongins A and B and their stereoisomers at C-5.
Kawai N, et al.
Tetrahedron, 63(37), 9049-9056 (2007)
Bis (oxazoline)-ligand-mediated asymmetric [2, 3]-Wittig rearrangement of benzyl ethers: reaction mechanism based on the hydrogen/deuterium exchange effect.
Kitamura M, et al.
Tetrahedron, 68(22), 4280-4285 (2012)

Articles

we are pleased to offer both enatiomers of 2-methyl-CBS-oxazaborolidine as a dry reagent, in addition to our current offerings as a 1M solution in toluene.

Learn about the applications of chiral oxazaborolidinium ions (COBIs) as Lewis acid catalysts in different asymmetric reactions such as cyclopropanation, epoxidation, and radical reactions along with details of their catalytic action.

Learn about the applications of chiral oxazaborolidinium ions (COBIs) as Lewis acid catalysts in different asymmetric reactions such as cyclopropanation, epoxidation, and radical reactions along with details of their catalytic action.

Learn about the applications of chiral oxazaborolidinium ions (COBIs) as Lewis acid catalysts in different asymmetric reactions such as cyclopropanation, epoxidation, and radical reactions along with details of their catalytic action.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service