Skip to Content
Merck
All Photos(2)

Key Documents

450162

Sigma-Aldrich

Potassium hexachloroiridate(IV)

99.99% trace metals basis

Synonym(s):

Iridium potassium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2IrCl6
CAS Number:
Molecular Weight:
483.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

>385 °C (lit.)

SMILES string

[K+].[K+].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.Ir.2K/h6*1H;;;/q;;;;;;+4;2*+1/p-6

InChI key

WEKZJZDULNNSCD-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Voltammetry at HMT-PMBI-coated glassy carbon electrodes: This study explores the electrochemical properties of potassium hexachloroiridate(IV) and its potential for sensitive detection, which could be valuable in analytical chemistry applications (M Rees et al., 2020).
  • Antioxidant Total Capacity Assay: Introduces a novel assaying method for antioxidant capacity using potassium hexachloroiridate(IV), which could have implications in pharmacology and drug discovery (Y Liu et al., 2024).
  • Mass Transfer in 3D-printed Electrolyzers: Examines the role of potassium hexachloroiridate(IV) in enhancing mass transfer in electrolytic systems, which is pivotal for chemical engineering and energy applications (SJC Weusten et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Song et al.
Inorganic chemistry, 47(24), 11458-11460 (2008-11-14)
One-electron oxidation of phenol, 2,4,6-trimethylphenol, and 2,6-dimethylphenol by [IrCl(6)](2-) in aqueous solution has a simple pH dependence, indicating slow bimolecular oxidation of ArOH and faster oxidation of ArO(-). H/D kinetic isotope effects as large as 3.5 for oxidation of ArOH
Fabrizio Testa et al.
Free radical biology & medicine, 51(8), 1567-1574 (2011-08-16)
Unlike superoxide dismutases (SODs), superoxide reductases (SORs) eliminate superoxide anion (O(2)(•-)) not through its dismutation, but via reduction to hydrogen peroxide (H(2)O(2)) in the presence of an electron donor. The microaerobic protist Giardia intestinalis, responsible for a common intestinal disease
Y Kaneko et al.
Biochemistry, 19(25), 5795-5799 (1980-12-09)
Zinc-substituted horseradish peroxidase is oxidized by K2IrCl6 to a characteristic state which retains one oxidizing equivalent more than the zinc peroxidase. The oxidized enzyme gives an optical absorption spectrum similar to that of compound I of peroxidase and catalase, and
Z S Farhangrazi et al.
Biochemistry, 34(46), 14970-14974 (1995-11-21)
Horseradish peroxidase (HRP) was oxidized by IrCl6(2-) to a mixture of compounds I and II, the rate of oxidation and the ratio of the mixture being greatly affected by pH (Hayashi & Yamazaki, 1979). Oxidation of HRP by IrCl6(2-) in
J G Muller et al.
Nucleic acids research, 26(9), 2247-2249 (1998-06-20)
Two gel electrophoretic methods are described for detection of 7, 8-dihydro-8-oxoguanine and 7,8-dihydro-8-oxoadenine based on their further oxidation with one-electron oxidants including IrCl62-and IrBr62-. The products of nucleobase oxidation lead to enhanced piperidine-sensitive cleavage and to highly visible stop points

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service