Skip to Content
Merck
All Photos(1)

Documents

448168

Sigma-Aldrich

Hexanoic-d11 acid

98 atom % D

Synonym(s):

Caproic acid-d11,OH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)4CO2H
CAS Number:
Molecular Weight:
127.23
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.412 (lit.)

bp

202-203 °C (lit.)

mp

-3 °C (lit.)

density

1.015 g/mL at 25 °C

mass shift

M+11

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3,2D2,3D2,4D2,5D2

InChI key

FUZZWVXGSFPDMH-GILSBCIXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth L Onderko et al.
Nature chemistry, 9(7), 623-628 (2017-06-24)
Strong electron-donation from the axial thiolate ligand of cytochrome P450 has been proposed to increase the reactivity of compound I with respect to C-H bond activation. However, it has proven difficult to test this hypothesis, and a direct link between
Liq. Cryst., 8, 31-31 (1990)
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service