Skip to Content
Merck
All Photos(2)

Key Documents

445711

Sigma-Aldrich

Poly(3-octylthiophene-2,5-diyl)

regioregular

Synonym(s):

P3OT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18S)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

mol wt

average Mn ~34,000

color

black

conductivity

1 × 10-6 S/cm
~1 × 103 S/cm (when doped with iodine)

mp

190 °C
198-211 °C

fluorescence

λex 442 nm; λem 562 nm in chloroform

OPV Device Performance

ITO/PEDOT:PSS/P3OT:PC61BM (1:2)/LiF/Al

  • Short-circuit current density (Jsc): 5.55 mA/cm2
  • Open-circuit voltage (Voc): 0.5 V
  • Fill Factor (FF): 0.33
  • Power Conversion Efficiency (PCE): 0.91 %

semiconductor properties

P-type (mobility=1E-4-1E-1)

SMILES string

[s]1cc(cc1)CCCCCCCC

InChI key

WQYWXQCOYRZFAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Greater than 98.5% head-to-tail regiospecific conformation.
Poly(3-octylthiophene-2,5-diyl) (P3OT) is an alkylthiophene based conductive polymer with high electroluminescence, which is soluble in most of the organic solvents. It can be prepared by the electrochemical polymerization of 3-octylthiophene (3OT). It is hydrophobic and can be used as a transducer for ion selective electrodes.

Application

For the characterization and solid-state properties of this polymer, see J. Am. Chem. Soc. .
P3OT forms a nanocomposite with carbon nanotubes (CNTs) for devising a high performance semiconducting material. It can also be used as a π-conjugating polymer for the fabrication of organic electronic based devices which include polymeric diodes, solar cells and photovoltaic modules.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials. Poly (3-octylthiophene-2,5-diyl) (P3OT)-single walled carbon nanotubes composites were prepared., 3 Doped P3OT may be used in NO2 sensing applications. 2
Conducting polymer.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
Used in organic field-effect transistors and in polymer-based solar cells.

Features and Benefits

Good processability, environmental stability and electroactivity.

Packaging

Packaged in glass bottles

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical characterizations of semi-conducting conjugated polymer-CNTs nanocomposites.
Abu-Abdeen M, et al.
Journal of Polymer Research, 19(3), 1-9 (2012)
Poly (3-octylthiophene) as solid contact for ion-selective electrodes: contradictions and possibilities
Jarvis JM, et al.
Journal of Solid State Electrochemistry, 20(11), 3033-3041 (2016)
Semiconducting polymer diodes: Large size, low cost photodetectors with excellent visible-ultraviolet sensitivity
Yu, G and Pakbaz, K and Heeger, AJ
Applied Physics Letters, 64(25), 3422-3424 (1994)
Fabrication of a poly (3-octylthiophene-2, 5-diyl) electrochemiluminescence device assisted by perylene
Daimon T and Nihei E
Materials, 6(5), 1704-1717 (2013)
Effects of annealing on the nanomorphology and performance of poly (alkylthiophene): fullerene bulk-heterojunction solar cells
Nguyen LH, et al.
Advances in Functional Materials, 17(7), 1071-1078 (2007)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service