Skip to Content
Merck
All Photos(1)

Key Documents

442194

Sigma-Aldrich

3-Methoxyphenylmagnesium bromide solution

1.0 M in THF

Synonym(s):

(m-Methoxyphenyl)magnesium bromide, 3-Anisylmagnesium bromide, m-Anisylmagnesium bromide, Bromo(3-methoxyphenyl)magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4MgBr
CAS Number:
Molecular Weight:
211.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

bp

65-67 °C

density

1.013 g/mL at 25 °C

SMILES string

COc1cccc([Mg]Br)c1

InChI

1S/C7H7O.BrH.Mg/c1-8-7-5-3-2-4-6-7;;/h2-3,5-6H,1H3;1H;/q;;+1/p-1

InChI key

AJKNYEHIJAMGHX-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service