Skip to Content
Merck
All Photos(1)

Key Documents

440736

Sigma-Aldrich

4-Thiouracil

97%

Synonym(s):

2-Hydroxy-4-mercaptopyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2OS
CAS Number:
Molecular Weight:
128.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

295 °C (dec.) (lit.)

solubility

1 M NaOH: soluble 50 mg/mL

SMILES string

Oc1nccc(S)n1

InChI

1S/C4H4N2OS/c7-4-5-2-1-3(8)6-4/h1-2H,(H2,5,6,7,8)

InChI key

OVONXEQGWXGFJD-UHFFFAOYSA-N

Application

4-Thiouracil is suitable reagent employed in Schneider′s media for the embryos during RNA extraction. It may be employed as reagent for the Northwestern blotting technique.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and tautomerism of the neutral and monoanionic forms of 4-thiouracil derivatives.
A Psoda et al.
Journal of the American Chemical Society, 96(22), 6832-6839 (1974-10-30)
Jeanine S Morey et al.
PloS one, 8(6), e66347-e66347 (2013-06-19)
Dinoflagellates possess many physiological processes that appear to be under post-transcriptional control. However, the extent to which their genes are regulated post-transcriptionally remains unresolved. To gain insight into the roles of differential mRNA stability and de novo transcription in dinoflagellates
Michael R Miller et al.
Nature methods, 6(6), 439-441 (2009-05-12)
We found that the combination of spatially restricted uracil phosphoribosyltransferase (UPRT) expression with 4-thiouracil delivery can be used to label and purify cell type-specific RNA from intact complex tissues in Drosophila melanogaster. This method is useful for isolating RNA from
Margaret A Nakamoto et al.
RNA (New York, N.Y.), 23(12), 1834-1849 (2017-08-31)
RNA contains over 100 modified nucleotides that are created post-transcriptionally, among which pseudouridine (Ψ) is one of the most abundant. Although it was one of the first modifications discovered, the biological role of this modification is still not fully understood.
Linda Warfield et al.
Molecular cell, 68(1), 118-129 (2017-09-19)
Previous studies suggested that expression of most yeast mRNAs is dominated by either transcription factor TFIID or SAGA. We re-examined the role of TFIID by rapid depletion of S. cerevisiae TFIID subunits and measurement of changes in nascent transcription. We find that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service