440728
2,5-Dimethyl-3-furoic acid
98%
Synonym(s):
2,5-Dimethyl-3-furancarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
137-140 °C (lit.)
functional group
carboxylic acid
SMILES string
Cc1cc(C(O)=O)c(C)o1
InChI
1S/C7H8O3/c1-4-3-6(7(8)9)5(2)10-4/h3H,1-2H3,(H,8,9)
InChI key
CNTHHNPBADVTRY-UHFFFAOYSA-N
Application
2,5-Dimethyl-3-furoic acid may be used as carboxylato ligand in the preparation of the following titanocene complexes:
- [Ti(η5-C5H5)2(OOC-dmf)2]
- [Ti(η5-C5H4Me)2(OOC-dmf)2]
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis, characterization and in vitro biological studies of titanocene (IV) derivatives containing different carboxylato ligands.
Ceballos-Torres J, et al.
Journal of Organometallic Chemistry, 716, 201-207 (2012)
Santiago Gómez-Ruiz et al.
Journal of inorganic biochemistry, 102(12), 2087-2096 (2008-09-02)
The reaction of 3-methoxyphenylacetic acid (3-MPAH), 4-methoxyphenylacetic acid (4-MPAH), 2,5-dimethyl-3-furoic acid (DMFUH) or 1,4-benzodioxane-6-carboxylic acid (BZDOH) with triphenyltin(IV) chloride (1:1) or diphenyltin(IV) dichloride (2:1) in the presence of triethylamine yielded the compounds [SnPh3(3-MPA)] (1), [SnPh3(4-MPA)] (2), [SnPh3(DMFU)] (3), [SnPh3(BZDO)] (4)
Lourdes Rocamora-Reverte et al.
ChemMedChem, 7(2), 301-310 (2011-12-16)
A group of organotin(IV) complexes were prepared: [SnCy3 (DMNI)] (1), [SnCy3 (BZDO)] (2), [SnCy3 (DMFU)] (3), and [SnPh2 (BZDO)2 ] (4), for which DMNIH=2,6-dimethoxynicotinic acid, BZDOH=1,4-benzodioxane-6-carboxylic acid, and DMFUH=2,5-dimethyl-3-furoic acid. The cytotoxic activities of compounds 1-4 were tested against pancreatic
Milena R Kaluderović et al.
European journal of medicinal chemistry, 45(2), 519-525 (2009-11-21)
The reaction of 3-methoxyphenylacetic acid, 4-methoxyphenylacetic acid, mesitylthioacetic acid, 2,5-dimethyl-3-furoic acid and 1,4-benzodioxane-6-carboxylic acid with trimethylgallium (1:1) yielded the dimeric complexes [Me(2)Ga(micro-O(2)CCH(2)C(6)H(4)-3-OMe)](2) (1), [Me(2)Ga(micro-O(2)CCH(2)C(6)H(4)-4-OMe)](2) (2), [Me(2)Ga(micro-O(2)CCH(2)SMes)](2) (3) (Mes=2,4,6-Me(3)C(6)H(2)), [Me(2)Ga{micro-O(2)C(Fur)}](2) (4) (Fur=2,5-dimethylfuran) and [Me(2)Ga{micro-O(2)C(Bdo)}](2) (5) (Bdo=1,4-benzodioxane) respectively. The molecular structure of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service