Skip to Content
Merck
All Photos(3)

Key Documents

434574

Sigma-Aldrich

Poly(sodium 4-styrenesulfonate)

average Mw ~1,000,000, powder

Synonym(s):

PSS, Poly(4-styrenesulfonic acid) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H7NaO3S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mw ~1,000,000

viscosity

70-170 cP, 10 wt. % in H2O(25 °C, Brookfield)(lit.)

mp

450 °C (dec.)

solubility

H2O: soluble

density

0.801 g/mL at 25 °C (lit.)

SMILES string

[S](=O)(=O)(O)c1ccc(cc1)C=C

InChI

1S/C8H8O3S/c1-2-7-3-5-8(6-4-7)12(9,10)11/h2-6H,1H2,(H,9,10,11)

InChI key

MAGFQRLKWCCTQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dispersant for metal oxides and sulfates, suspending agent for particulates, protective colloid for oil-in-water emulsions and viscosity modifier in aqueous-based adhesives.

Features and Benefits

Anionic polyelectrolyte.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jagadeesh K Venkatesan et al.
Tissue engineering. Part A, 26(7-8), 450-459 (2019-11-05)
Scaffold-guided gene transfer offers strong systems to develop noninvasive convenient therapeutic options for the treatment of articular cartilage defects, especially when targeting bone marrow aspirates from patients containing chondroregenerative mesenchymal stromal cells in a native microenvironment. In this study, we
Jagadeesh K Venkatesan et al.
Pharmaceutics, 12(3) (2020-04-05)
The delivery of therapeutic genes in sites of articular cartilage lesions using non-invasive, scaffold-guided gene therapy procedures is a promising approach to stimulate cartilage repair while protecting the cargos from detrimental immune responses, particularly when targeting chondroreparative bone marrow-derived mesenchymal
Eduardo Lanzagorta Lanzagorta Garcia et al.
Materials (Basel, Switzerland), 14(5) (2021-03-07)
An innovative antimicrobial technology for plastic surfaces is presented. We report the synthesis and scale-up of triangular silver nanoplates (TSNPs) and their integration into polycaprolactone (PCL) and polylactic acid (PLA) polymers through a solvent-casting technique. The TSNPs have a high
Karen M Gallegos et al.
Frontiers in microbiology, 9, 3269-3269 (2019-02-01)
Chlamydia is the most frequently reported sexually transmitted bacteria causing 2.9 million infections annually in the United States. Diagnosis, treatment, and sequelae of chlamydial disease cost billions of dollars each year in the United States alone. Considering that a heparin
Johannes Frueh et al.
Physical chemistry chemical physics : PCCP, 15(2), 483-488 (2012-11-23)
Polyelectrolyte multilayers (PEMs) deposited on flexible supports are promising candidates for many applications ranging from controlled wettability over stimuli responsive nanovalves to lithography free surface structuring. Since many potential applications involve elongation of these films, we investigated the effect of

Articles

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service