Skip to Content
Merck
All Photos(2)

Key Documents

434256

Sigma-Aldrich

2-(Chloromethyl)anthraquinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9ClO2
CAS Number:
Molecular Weight:
256.68
Beilstein:
1975355
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

166-169 °C (lit.)

functional group

chloro
ketone

SMILES string

ClCc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C15H9ClO2/c16-8-9-5-6-12-13(7-9)15(18)11-4-2-1-3-10(11)14(12)17/h1-7H,8H2

InChI key

NVUYDKYMEMGYFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Chloromethyl)anthraquinone is an anthraquinone derivative. It was utilized as a substrate to synthesize Cibanone yellow R, a vat dye. Its electrochemical behavior has been studied over a wide pH range.

Application

2-(Chloromethyl)anthraquinone may be used in the following processes:
  • Synthesis of 2-[methylamino-N-(1′-methyl-4′-N,N-diethylaminobutyl)]anthraquinone diphosphate.
  • Synthesis of 2-(3,4-dicyano-phenyl)-2-(9,10-dioxo-9,10-dihydroantracen-2-yl-methyl)-malonic acid diethyl ester, starting reagent for the synthesis of tetra substituted metallophthalocyanines.
  • To alter the glassy carbon (GC) electrode surface in order to investigate the oxygen reduction reaction (ORR) in alkaline medium.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulphur dyes and sulphurised vat dyes.
Shah KH, et al.
Proceedings of the Indian Academy of Sciences - Section A, 30(1) (1949)
Synthesis, electrochemistry, spectroelectrochemistry and electrocolorimetry of phthalocyanine-anthraquinone hybrids.
Sezer B, et al.
Synt. Metals, 160(19), 2155-2166 (2010)
Electrochemical investigations of unexplored anthraquinones and their DNA binding.
Shah A, et al.
Journal of Electrochemical Science and Engineering, 3(1), 19-27 (2013)
Electrocatalysis of oxygen reduction on glassy carbon electrodes modified with anthraquinone moieties.
Mooste M, et al.
Journal of Solid State Electrochemistry, 18(6), 1725-1733 (2014)
Kevin K Schrader et al.
Applied and environmental microbiology, 69(9), 5319-5327 (2003-09-06)
Musty "off-flavor" in pond-cultured channel catfish (Ictalurus punctatus) costs the catfish production industry in the United States at least 30 million US dollars annually. The cyanobacterium Oscillatoria perornata (Skuja) is credited with being the major cause of musty off-flavor in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service