Skip to Content
Merck
All Photos(1)

Documents

433632

Sigma-Aldrich

Hydrazine solution

1.0 M in THF

Synonym(s):

Hydrazine, Nitrogen hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2
CAS Number:
Molecular Weight:
32.05
Beilstein:
878137
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in THF

bp

65 °C

density

0.886 g/mL at 25 °C

SMILES string

NN

InChI

1S/H4N2/c1-2/h1-2H2

InChI key

OAKJQQAXSVQMHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrazine promotes the cation exchange reaction for fast doping Cu ions into ZnSe nanocrystals.

Application

Hydrazine solution may be employed for the following studies:
  • As reducing agent for the reduction of the first row (3d) transition metal complexes in aqueous solution.
  • Preparation of nanoscale reduced metal oxides such as VO2(B), Cr2O3.
  • As fuel for the direct hydrazine fuel cell (DHFC).
  • As reducing agent for the synthesis of metallic monodisperse copper nanoparticles.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tomasz Kruk et al.
Colloids and surfaces. B, Biointerfaces, 128, 17-22 (2015-02-28)
Metallic monodisperse copper nanoparticles at a relatively high concentration (300 ppm CuNPs) have been synthesized by the reduction of copper salt with hydrazine in the aqueous SDS solution. The average particles size and the distribution size were characterized by Dynamic
Haibao Shao et al.
Journal of fluorescence, 25(2), 305-310 (2015-01-22)
Controllable doping is an effective way of tuning the properties of semiconductor nanocrystals (NCs). In this work, a simple strategy of fast doping Cu ions into ZnSe NCs under ambient conditions was proposed. The principle of doping is based on
Synthesis and characterization of reduced transition metal oxides and nanophase metals with hydrazine in aqueous solution.
Gui Z, et al.
Materials Research Bulletin, 38(1), 169-176 (2003)
Effects of NaOH addition on performance of the direct hydrazine fuel cell.
Yin WX, et al.
Journal of Power Sources, 182(2), 520-523 (2008)
M Vogel et al.
Fresenius' journal of analytical chemistry, 366(8), 781-791 (2001-03-03)
Hydrazine reagents are a well-known group of derivatizing agents for the determination of aldehydes and ketones in liquid and gaseous samples. Within this article, the most important hydrazine reagents are critically summarized, and their major applications in different fields, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service