Skip to Content
Merck
All Photos(3)

Documents

431648

Sigma-Aldrich

Sodium sulfide nonahydrate

≥99.99% trace metals basis

Synonym(s):

Disodium sulfide nonahydrate, Sodium sulfide enneahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S·9H2O
CAS Number:
Molecular Weight:
240.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: sodium

impurities

<100 ppm total metallic impurities

anion traces

SO32- and S2O32-: ≤0.1%

cation traces

Fe:, passes test
NH4+: ≤0.005%

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.O.O.[Na]S[Na]

InChI

1S/2Na.9H2O.S/h;;9*1H2;

InChI key

UIINQVYGFKNSPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfide nonahydrate is a chemical compound with the formula Na2S-H2O. It is a white to yellow hygroscopic solid that is highly soluble in water and has a strong, unpleasant odor. Sodium sulfide nonahydrate is typically made by dissolving elemental sulfur in a solution of sodium hydroxide, which leads to the formation of sodium sulfide. The nonahydrate form of the compound is formed by adding excess water to the solution, which leads to the formation of a crystalline solid with nine molecules of water per molecule of sodium sulfide. Sodium sulfide nonahydrate is often used as a reducing agent in chemical reactions as well as a flotation agent in the mining industry. It is also used in the production of dyes, pigments, and other chemicals.

Application

Sodium sulfide nonahydrate (Na2S.9H2O) can be reacted with α,ω-dibromo and/or α,ω-dichloroalkane for the synthesis of polyalkylene sulfide. It can be used as a reducing agent for the removal of nitrogen oxide from the exhaust gas. Na2S.9H2O can also be used as an inexpensive sulfur source for the synthesis of heterocyclic thiochromes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Absorption-reduction technique assisted by ozone injection and sodium sulfide for NOx removal from exhaust gas
Mok YS
Chemical Engineering Journal, 118(1-2), 63-67 (2006)
The synthesis of polymeric sulfides by reaction of dihaloalkanes with sodium sulfide
Smith K, et al.
Journal of Sulfur Chemistry, 32(6), 521-531 (2011)
The reaction products of sulfide and S-nitrosoglutathione are potent vasorelaxants.
Berenyiova A, et al.
Nitric Oxide, 46, 123-130 (2015)
Convenient one-pot access to 2 H-3-nitrothiochromenes from 2-bromobenzaldehydes, sodium sulfide and beta-nitrostyrenes
Le TTH, et al.
Organic & Biomolecular Chemistry, 17(26), 6355-6358 (2019)
Veinot, J.G.C. et al.
Chemistry of Materials, 9, 2117-2117 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service