Skip to Content
Merck
All Photos(1)

Documents

42358

Sigma-Aldrich

Dioctadecylamine

≥99.0% (NT)

Synonym(s):

DODA, Distearylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)17]2NH
CAS Number:
Molecular Weight:
521.99
Beilstein:
1801688
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

mp

71-73 °C

solubility

water: soluble(lit.)

SMILES string

CCCCCCCCCCCCCCCCCCNCCCCCCCCCCCCCCCCCC

InChI

1S/C36H75N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37H,3-36H2,1-2H3

InChI key

HKUFIYBZNQSHQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dioctadecylamine (DODA), a secondary amine, is a fatty amine derivative. Its biodegradation potential has been assessed. It has been observed that in aqueous medium DODA self organizes into plate like structures. Its partial charge distribution as a function of its conformation has been analyzed. The phase diagram of dioctadecylamine Langmuir monolayers has been determined and investigations show it does not form a monolayer above pH 3.9. DODA has been utilized in forming liposomes, cationic lipids or lipid chelating agents.

Application

Dioctadecylamine (DODA) is suitable reagent used in the synthesis of the following:
  • Dioctadecylamine-BCN (bicyclo[6.1.0]nonyne) conjugate.
  • Lipid derivatives of bisethylnorspermine (BSP).
  • Functional VP (N-vinylpyrrolidone ) polymers.
It may be used in the following studies:
  • As a reactant in the synthesis of 4,4′-azobis(4-cyano-N,N-dioctadecyl)pentanamide (DODA-501) by reacting with disuccinimidyl 4,4′-azobis(4-cyanovalerate).
  • As a reagent in the synthesis of dioctadecyl heptapeptides.
  • As a phase transfer and stabilizer agent for gold nanoparticles (AuNPs) in non-polar solvent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The dioctadecylamine monolayer: Textures, phase transitions, and dendritic growth.
Flores A, et al.
J. Chem. Phys. , 119(11), 5644-5653 (2003)
Samuel F Brockington et al.
The New phytologist, 207(4), 1170-1180 (2015-05-15)
Betalain pigments are unique to the Caryophyllales and structurally and biosynthetically distinct from anthocyanins. Two key enzymes within the betalain synthesis pathway have been identified: 4,5-dioxygenase (DODA) that catalyzes the formation of betalamic acid and CYP76AD1, a cytochrome P450 gene
Emmanuelle Roux et al.
Journal of pharmaceutical sciences, 91(8), 1795-1802 (2002-07-13)
The aim of this study was to characterize a pH-sensitive liposome formulation bearing a terminally alkylated N-isopropylacrylamide (NIPAM) copolymer with regard to its pH responsiveness, surface properties, and pharmacokinetics. The interacting forces between two lipid bilayers bearing the anchored NIPAM
Glycosidation of alkylamino-alkan-1-ol. A simple and convenient synthesis of glycosylated cationic lipids.
Jacopin C, et al.
Bioorganic & Medicinal Chemistry, 12(11), 1447-1450 (2002)
K Tagawa et al.
Bioconjugate chemistry, 10(3), 354-360 (1999-05-29)
Novel amphiphiles which carry many mannose residues as side chains were prepared by telomerization of N-methacryloylaminopropyl D-mannopyranoside (alpha:beta = 20:1), N-methacryloylaminohexyl D-mannopyranoside (alpha:beta = 20:1), or 3-(2-methacryloylaminoethylthio)propyl D-mannopyranoside (alpha:beta = 4:1) using a lipophilic radical initiator. The mannose-carrying amphiphiles incorporated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service