Skip to Content
Merck
All Photos(2)

Key Documents

419036

Sigma-Aldrich

Cellulose acetate butyrate

average Mn ~12,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn ~12,000

extent of labeling

16-19 wt. % Acetyl
30-35 wt. % Butyryl

refractive index

n20/D 1.475 (lit.)

density

1.25 g/mL at 25 °C (lit.)

SMILES string

O5[C@H]([C@@H]([C@H]([C@@H]([C@H]5COC(=O)CCC)OC(=O)CCC)OC(=O)CCC)OC(=O)CCC)O[C@@H]6[C@H](O[C@H]([C@@H]([C@H]6OC(=O)CCC)OC(=O)CCC)OC(=O)CCC)COC(=O)CCC.O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OC(=

InChI

1S/C44H70O19.C28H38O19.C12H22O11/c1-9-17-29(45)53-25-27-37(57-31(47)19-11-3)39(58-32(48)20-12-4)42(61-35(51)23-15-7)44(56-27)63-38-28(26-54-30(46)18-10-2)55-43(62-36(52)24-16-8)41(60-34(50)22-14-6)40(38)59-33(49)21-13-5;1-11(29)37-9-19-21(39-13(3)31)23(40

InChI key

WGKMZYZRLDSCLX-OLZTYWLQSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitra Jelvehgari et al.
Archives of pharmacal research, 32(7), 1019-1028 (2009-07-31)
The present research work compares the effect of microsphere preparation technique on micromeritics and release behaviors of theophylline microspheres. Microspheres were prepared by oil-in oil (O(1)/O(2)) emulsion solvent evaporation method (ESE) using different ratios of anhydrous theophylline to cellulose acetate
Soad A Yehia et al.
Drug delivery, 18(8), 620-630 (2011-11-25)
The purpose of this study is to increase the lag time and prevent release of budesonide, a corticosteroid drug used in Crohn's disease for the first 5 h and efficiently deliver it to the colon. Eudragit S100 spray-coated capsules and pulsatile
M Constantin et al.
International journal of pharmaceutics, 330(1-2), 129-137 (2006-10-10)
Poly(vinyl alcohol) (PVA) microspheres were prepared by dispersion reticulation with glutaraldehyde and further aminated. These microspheres were firstly loaded with diclofenac (DF) and then entrapped in cellulose acetate butyrate (CAB) microcapsules by an o/w solvent evaporation technique for intestinal delivery
Varaporn Buraphacheep Junyaprasert et al.
International journal of pharmaceutics, 352(1-2), 81-91 (2007-12-07)
A sustained release suspension of diltiazem, a short half-life calcium channel blocker, was developed to reduce frequency of drug administration, ease of dose adjustment and improve patient compliance. In this study, the sustained release of diltiazem was obtained by complexing
Chenyang Xing et al.
Carbohydrate polymers, 92(2), 1921-1927 (2013-02-13)
The eco-friendly poly(propylene carbonate) (PPC)/cellulose acetate butyrate (CAB) blends were prepared by melt-blending in a batch mixer for the first time. PPC and CAB were partially miscible because of the drastically shifted glass transition temperatures of both PPC and CAB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service