Skip to Content
Merck
All Photos(1)

Key Documents

418102

Sigma-Aldrich

Butyraldehyde

purified by redistillation, ≥99.5%

Synonym(s):

Butanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CHO
CAS Number:
Molecular Weight:
72.11
Beilstein:
506061
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.5 (vs air)

Quality Level

vapor pressure

90 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

390 °F

purified by

redistillation

expl. lim.

12.5 %

refractive index

n20/D 1.380 (lit.)

bp

75 °C (lit.)

mp

−96 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

functional group

aldehyde

Organoleptic

pungent

SMILES string

[H]C(=O)CCC

InChI

1S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3

InChI key

ZTQSAGDEMFDKMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyraldehyde, also known as Butanal, is an aliphatic aldehyde, that is commonly used as a versatile building block in various organic reactions, such as in the synthesis of diverse C4–C8 alcohols, esters, amines, and carboxylic acids.

Application

Butyraldehyde can be used as a reactant in the:
  • Reductive amination of various nitroarene derivatives to synthesize mono-N-alkylated aniline derivatives.
  • Ionic liquid-catalyzed Knoevenagel condensation with active methylene compounds to synthesize substituted electrophilic alkenes.
  • NiCl2-catalyzed synthesis of α-aminonitriles by reacting with amines and trimethylsilyl cyanides.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<50.0 °F - Pensky-Martens closed cup

Flash Point(C)

< 10 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The condensation of butyraldehyde and aniline.
Kharasch MS, et al.
Journal of the American Chemical Society, 62(3), 494-497 (1940)
Heterogeneous photocatalytic oxidation of gas-phase organics for air purification: acetone, 1-butanol, butyraldehyde, formaldehyde, and m-xylene oxidation.
Peral J and Ollis DF.
J. Catal., 1136(2), 554-565 (1992)
Phosphazene base promoted anionic polymerization of n-butyraldehyde
Inge W, et al.
European Polymer Journal, 93, 97-102 (2017)
Aldol addition of butyraldehyde over solid base catalysts.
Zhang G, et al.
Bulletin of the Chemical Society of Japan, 62(6), 2070-2072 (1989)
Self-condensation of n-butyraldehyde over solid base catalysts.
Tsuji H, et al.
J. Catal., 148(2), 759-770 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service