Skip to Content
Merck
All Photos(1)

Documents

414476

Sigma-Aldrich

Ethyl 3,3-diethoxypropionate

90%, technical grade

Synonym(s):

Malonaldehydic acid ethyl ester diethylacetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CHCH2CO2C2H5
CAS Number:
Molecular Weight:
190.24
Beilstein:
1772110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

92-93 °C/13 mmHg (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

CCOC(CC(=O)OCC)OCC

InChI

1S/C9H18O4/c1-4-11-8(10)7-9(12-5-2)13-6-3/h9H,4-7H2,1-3H3

InChI key

SIALOQYKFQEKOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 3,3-diethoxypropionate participates in the Yb(OTf)3 catalyzed synthesis of dihydropyridines (DHPs).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ram Reddy et al.
European journal of medicinal chemistry, 62, 395-404 (2013-02-06)
Montmorillonite K-10 mediated MCR of anilines, arylaldehydes and ethyl-3,3-diethoxypropionate in water afforded 2,6-unsubstituted dihydropyridines depending on the nature of anilines employed. A variety of dihydropyridines were prepared by using this green methodology in good yields and montmorillonite K-10 was found
Construction of a quinoline ring via a 3-component reaction in water: crystal structure analysis and H-bonding patterns of a 2-aryl quinoline.
Reddy TR, et al.
Green Chemistry, 14(7), 1870-1872 (2012)
Ytterbium-catalyzed synthesis of dihydropyridines.
Sueki S, et al.
Tetrahedron Letters, 52(34), 4473-4477 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service