Skip to Content
Merck
All Photos(3)

Documents

409758

Sigma-Aldrich

N,N-Dimethyl-1,4-phenylenediamine oxalate

98%

Synonym(s):

N,N-Dimethyl-p-phenylenediamine hemioxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NC6H4NH2]2·H2C2O4
CAS Number:
Molecular Weight:
362.42
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

205 °C (dec.) (lit.)

SMILES string

OC(=O)C(O)=O.CN(C)c1ccc(N)cc1.CN(C)c2ccc(N)cc2

InChI

1S/2C8H12N2.C2H2O4/c2*1-10(2)8-5-3-7(9)4-6-8;3-1(4)2(5)6/h2*3-6H,9H2,1-2H3;(H,3,4)(H,5,6)

InChI key

MNUINXKPLPIOEF-UHFFFAOYSA-N

Application

N,N-Dimethyl-1,4-phenylenediamine oxalate (N,N-dimethyl-p-phenylenediamine hemioxalate salt, DPD) was used as reagent in a study to evaluate the catalytic effects of elemental sulfur nanoparticles on CrVI reduction. It was also used to study the transformation of trichloroethene (TCE), caused by heating contaminated soil and groundwater samples.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On-line electrochemical-mass spectrometry study of the mechanism of oxidation of N, N-dimethyl-p-phenylenediamine in aqueous electrolytes.
Modestov AD, et al.
Journal of Electroanalytical Chemistry, 565(1), 7-19 (2004)
Jed Costanza et al.
Environmental science & technology, 43(3), 909-914 (2009-02-28)
This study explores the transformation of trichloroethene (TCE) caused by heating contaminated soil and groundwater samples obtained from the East Gate Disposal Yard (EGDY) located in Fort Lewis, WA. After field samples transferring into glass ampules and introducing 1.5 micromol
Yeqing Lan et al.
Environmental science & technology, 39(7), 2087-2094 (2005-05-06)
Chromate (CrVI) reduction by sulfide was conducted in anaerobic batch experimental systems. The molar ratio of the reduced CrVI to the oxidized S(-II) was 1:1.5 during the reaction, suggesting that the product of sulfide oxidation was elemental sulfur. Under the
V Fogliano et al.
Journal of agricultural and food chemistry, 47(3), 1035-1040 (1999-12-20)
A novel method for measuring the antioxidant activity using N, N-dimethyl-p-phenylenediamine (DMPD) was developed. The radical cation of this compound gives a stable colored solution and a linear inhibition of color formation can be observed in the presence of 0.
Francisco J Morales et al.
Journal of agricultural and food chemistry, 50(16), 4657-4661 (2002-07-26)
The Maillard reaction has a dramatic impact on the overall acceptance and nutritional quality of most of the foods consumed daily in European countries. Melanoidins are polymeric structures formed in the last stage of the Maillard reaction with nearly unknown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service