Skip to Content
Merck
All Photos(1)

Key Documents

409359

Sigma-Aldrich

Scandium(III) chloride

anhydrous, powder, 99.9% trace metals basis

Synonym(s):

Scandium chloride, Scandium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ScCl3
CAS Number:
Molecular Weight:
151.31
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: scandium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

960 °C (lit.)

density

2.39 g/mL at 25 °C (lit.)

SMILES string

Cl[Sc](Cl)Cl

InChI

1S/3ClH.Sc/h3*1H;/q;;;+3/p-3

InChI key

DVMZCYSFPFUKKE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Scandium(III) chloride (ScCl3) is a scandium halide that can be formed by dissolving scandium oxide in an acid solution and evaporating the resultant solution to dryness.[1]

Application

ScCl3 can be used in the preparation of lithium salt of anionic scandium tetraborohydride complex (LiSc(BH4)4) by forming a ball-milled mixture. LiSc(BH4)4 can be used as a potential hydrogen storage material.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and mechanism of formation of anhydrous scandium (III) chloride and bromide
Stotz RW and Melson GA
Inorganic Chemistry, 11(7), 1720-1721 (1972)
LiSc (BH4) 4 as a hydrogen storage material: multinuclear high-Resolution solid-state NMR and first-Principles Density functional Theory Studies
Kim C, et al.
The Journal of Physical Chemistry C, 113(22), 9956-9968 (2009)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service