Skip to Content
Merck
All Photos(1)

Documents

404322

Sigma-Aldrich

Dichloro(1,2-diaminocyclohexane)platinum(II)

Synonym(s):

(1,2-Diaminocyclohexane)platinum(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10(NH2)2]PtCl2
CAS Number:
Molecular Weight:
380.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: platinum
reagent type: catalyst

mp

280 °C (dec.) (lit.)

SMILES string

Cl[Pt]Cl.NC1CCCCC1N

InChI

1S/C6H14N2.2ClH.Pt/c7-5-3-1-2-4-6(5)8;;;/h5-6H,1-4,7-8H2;2*1H;/q;;;+2/p-2

InChI key

PNNCIXRVXCLADM-UHFFFAOYSA-L

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Lact. - Muta. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachiko Kaida et al.
Cancer research, 70(18), 7031-7041 (2010-08-06)
Nanoparticle therapeutics are promising platforms for cancer therapy. However, it remains a formidable challenge to assess their distribution and clinical efficacy for therapeutic applications. Here, by using multifunctional polymeric micellar nanocarriers incorporating clinically approved gadolinium (Gd)-based magnetic resonance imaging contrast
A platinum(IV) complex oxidizes guanine to 8-oxo-guanine in DNA and RNA.
S Choi et al.
Inorganic chemistry, 40(22), 5481-5482 (2001-10-16)
L M Ruiz-Perez et al.
Tropical medicine and parasitology : official organ of Deutsche Tropenmedizinische Gesellschaft and of Deutsche Gesellschaft fur Technische Zusammenarbeit (GTZ), 38(1), 45-48 (1987-03-01)
The trypanocidal activity of three new compounds derived of the antineoplastic drug cis-diamminodichloroplatinum(II) (DDP) has been studied over epimastigote forms of Trypanosoma cruzi. The electron microscopy indicated that, in the treated animals, the nuclear chromatin is distributed in the nucleus
Kensuke Osada et al.
Journal of the American Chemical Society, 134(32), 13172-13175 (2012-07-28)
Spontaneous formation of polymeric metallosomes with uniform size (~100 nm) was found to occur in aqueous medium through the reaction of an anticancer agent, (1,2-diaminocyclohexane)platinum(II) (DACHPt), with a Y-shaped block copolymer of ω-cholesteroyl-poly(L-glutamic acid) and two-armed poly(ethylene glycol) (PEGasus-PLGA-Chole). Circular
S K Mauldin et al.
Cancer research, 48(18), 5136-5144 (1988-09-15)
We have previously reported the development of a two-column high performance liquid chromatography system for separation of platinum(II) complexes with the 1,2-diaminocyclohexane (DACH) carrier ligand (Mauldin et al., Anal. Biochem., 157: 129, 1986). Here we report the application of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service