Skip to Content
Merck
All Photos(3)

Documents

401285

Sigma-Aldrich

Cobalt(II) oxalate dihydrate

Synonym(s):

Cobalt(2+) oxalate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoC2O4 · 2H2O
CAS Number:
Molecular Weight:
182.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

concentration

29-35.4% Co (with A-Nitroso-B-Naphthol, gravimetric)

mp

230 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O.O.O=C1O[Co]OC1=O

InChI

1S/C2H2O4.Co.2H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;2*1H2/q;+2;;/p-2

InChI key

MWHSMSAKVHVSAS-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service