Skip to Content
Merck
All Photos(2)

Key Documents

392995

Sigma-Aldrich

Coumarin 314

Dye content 97 %

Synonym(s):

2,3,5,6-1H,4H-Tetrahydro-9-carbethoxyquinolizino-[9,9a,1-gh]coumarin, Coumarin 504

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

solid

composition

Dye content, 97%

mp

140-144 °C (lit.)
146.22-147.36 °C (lit.)

λmax

436 nm
437 nm in ethanol

ε (extinction coefficient)

≥39000 at 389-401 nm in ethanol at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCOC(=O)C1=Cc2cc3CCCN4CCCc(c2OC1=O)c34

InChI

1S/C18H19NO4/c1-2-22-17(20)14-10-12-9-11-5-3-7-19-8-4-6-13(15(11)19)16(12)23-18(14)21/h9-10H,2-8H2,1H3

InChI key

VMJKUPWQKZFFCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Laser dye.
Suitable as a laser dye

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason V Gregory et al.
Advanced healthcare materials, 9(21), e2000564-e2000564 (2020-09-23)
Delivery of multiple therapeutics has become a preferred method of treating cancer, albeit differences in the biodistribution and pharmacokinetic profiles of individual drugs pose challenges in effectively delivering synergistic drug combinations to and at the tumor site. Here, bicompartmental Janus
Seok Chan Park et al.
Polymers, 11(2) (2019-04-10)
In this study we present polymeric microneedles composed of multiple layers to control drug release kinetics. Layered microneedles were fabricated by spraying poly(lactic-co-glycolic acid) (PLGA) and polyvinylpyrrolidone (PVP) in sequence, and were characterized by mechanical testing and ex vivo skin
Andrea Doni et al.
The Journal of experimental medicine, 212(6), 905-925 (2015-05-13)
Pentraxin 3 (PTX3) is a fluid-phase pattern recognition molecule and a key component of the humoral arm of innate immunity. In four different models of tissue damage in mice, PTX3 deficiency was associated with increased fibrin deposition and persistence, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service