Skip to Content
Merck
All Photos(5)

Key Documents

392731

Sigma-Aldrich

(DHQD)2PHAL

greener alternative

≥95%

Synonym(s):

Hydroquinidine 1,4-phthalazinediyl diether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H54N6O4
CAS Number:
Molecular Weight:
778.98
Beilstein:
5475678
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

optical activity

[α]22/D −262°, c = 1.2 in methanol

greener alternative product score

old score: 5
new score: 3
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

160 °C (dec.) (lit.)

greener alternative category

SMILES string

CC[C@H]1CN2CCC1CC2[C@@H](Oc3nnc(O[C@H](C4CC5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c8ccccc38)c9ccnc%10ccc(OC)cc9%10

InChI

1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30-,31-,32-,43+,44+,45-,46-/m0/s1

InChI key

YUCBLVFHJWOYDN-HVLQGHBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(DHQD)2PHAL is a dimer that can be used as a chiral amine catalyst for asymmetric halogenations.

(DHQD)2PHAL is a modified cinchona alkaloid derivative mainly used as a ligand for enantioselective catalysis.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Less Hazardous Chemical Syntheses”, "Safer Solvents and Auxiliaries" and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

(DHQD)2PHAL may be used as ligand for:
  • Osmium trioxide catalyzed asymmetric dihydroxylation of olefins.
  • Palladium catalyzed Suzuki-Miyaura coupling of aryl/heteroaryl halides with aryl boronic acids in aqueous medium and in the absence of phosphine/organic solvent.
  • Copper(I)-catalyzed azide-alkyne cycloaddition reaction to synthesize 1,2,3-triazoles in water.
Hydroquinidine 1,4-phthalazinediyl diether [(DHQD)2PHAL] can be used:
  • As a ligand for Sharpless asymmetric dihydroxylation.
  • For enantioselective α-fluorination of carbonyl compounds.

Legal Information

Sold under license from Rhodia Pharma Solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(DHQD) 2 PHAL ligand-accelerated Cu-catalyzed azide-alkyne cycloaddition reactions in water at room temperature.
Ali AA, et al.
Royal Society of Chemistry Advances, 4(110), 64388-64392 (2014)
Syntheses and crystal structures of the cinchona alkaloid derivatives used as ligands in the osmium-catalyzed asymmetric dihydroxylation of olefins.
Amberg W, et al.
The Journal of Organic Chemistry, 58(4), 844-849 (1993)
Pd (OAc) 2 and (DHQD) 2 PHAL as a simple, efficient and recyclable/reusable catalyst system for Suzuki-Miyaura cross-coupling reactions in H 2 O at room temperature.
Saikia B, et al.
New. J. Chem., 39(4), 2440-2443 (2015)
Monomeric Cinchona Alkaloid-Based Catalysts for Highly Enantioselective Bromolactonisation of Alkynes
Wilking M, et al.
Chemistry?A European Journal , 22, 18601-18607 (2016)
Synthesis of enantiomerically enriched Baylis--Hillman alcohols from their acetates: combination of kinetic resolution during the salt formation with (DHQD) 2PHAL and following asymmetric induction during hydrolysis with NaHCO3 as a water surrogate
Kim J, et al.
Tetrahedron Letters, 43, 9141-9146 (2002)

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service