Skip to Content
Merck
All Photos(3)

Documents

385638

Sigma-Aldrich

Di-(2-picolyl)amine

97%

Synonym(s):

Bis(2-pyridylmethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3
CAS Number:
Molecular Weight:
199.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.578 (lit.)

bp

139-141 °C/1 mmHg (lit.)

solubility

water: soluble (partially miscible)(lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

C(NCc1ccccn1)c2ccccn2

InChI

1S/C12H13N3/c1-3-7-14-11(5-1)9-13-10-12-6-2-4-8-15-12/h1-8,13H,9-10H2

InChI key

KXZQYLBVMZGIKC-UHFFFAOYSA-N

Application

Di-(2-picolyl)amine can be used as:
  • A reactant to synthesize di(2-pyridylmethylamino)alkanoic esters using ω-iodoaliphatic carboxylic esters in the presence of triethylamine.
  • A ligand to synthesize iron complexes of di-(2-picolyl)amine by treating with iron trichloride, which can be used to catalyze the direct hydroxylation of benzene to phenol.
  • A reagent for the protection of carboxylic acids as carboxamides of bispicolylamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaohui Xu et al.
Materials (Basel, Switzerland), 13(17) (2020-08-23)
Fe3O4@SiO2 nanospheres with a core-shell structure were synthesized and functionalized with bis(2-pyridylmethyl)amine (BPMA). The photoresponses of the as-obtained Fe3O4@SiO2-BPMA for Cr3+, Cd2+, Hg2+ and Pb2+ ions were evaluated through irradiation with a 352 nm ultraviolet lamp, and Fe3O4@SiO2-BPMA exhibited remarkable
M Stylianou et al.
Dalton transactions (Cambridge, England : 2003), 46(11), 3688-3699 (2017-03-04)
A new two-electron photosensitizer, H
H M Dhammika Bandara et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(14), 3932-3941 (2011-03-03)
Photocages have been used to elucidate the biological functions of various small molecules and Ca(2+) ; however, there are very few photocages available for other metal ions. ZinCleav-2 (1-(4,5-dimethoxy-2-nitrophenyl)-N,N,N',N'-tetrakis-pyridin-2-ylmethyl-ethane-1,2-diamine) is a second-generation photocage for Zn(2+) that releases the metal ion
Bishnu Das et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(65), 14987-14995 (2020-08-28)
In this study we have analysed the comparative photophysical and electrochemical properties of two isomeric heterotrinuclear PtII -IrIII -PtII complexes 3 and 6 and the four corresponding intermediate isomeric homonuclear cyclometalated iridium(III) complexes 1, 2, 4 and 5. The isomerisation
Aimi Zhang et al.
Molecular imaging and biology, 22(3), 515-525 (2019-06-30)
This study aimed to synthesize a necrosis-avid agent using rhein as a precursor and labeled with gallium-68 (Ga-68) for positron emission tomography/computed tomography (PET/CT) imaging, to evaluate response to anticancer treatment in a mouse model. 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTA)-conjugated rhein was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service