Skip to Content
Merck
All Photos(2)

Key Documents

379883

Sigma-Aldrich

Nickel(II) acetate tetrahydrate

99.995% trace metals basis

Synonym(s):

Acetic acid nickel(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(OCOCH3)2 · 4H2O
CAS Number:
Molecular Weight:
248.84
Beilstein:
3730764
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.995% trace metals basis

form

powder, crystals or chunks

reaction suitability

core: nickel

density

1.798 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Ni]OC(C)=O

InChI

1S/2C2H4O2.Ni.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

OINIXPNQKAZCRL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clemente-Juan, J.M. et al.
Inorganic Chemistry, 38, 55-55 (1999)
Elham S Aazam et al.
Bioorganic chemistry, 57, 5-12 (2014-08-28)
Transition metal complexes compounds with Schiff bases ligand representing an important class of compounds that could be used to develop new metal-based anticancer agents and as precursors of metal NPs. Herein, 2,3-bis-[(3-ethoxy-2-hydroxybenzylidene)amino]but-2-enedinitrile Schiff base ligand and its corresponding copper/nickel complexes
Rusi et al.
PloS one, 10(7), e0129780-e0129780 (2015-07-15)
The composite metal oxide electrode films were fabricated using ex situ electrodeposition method with further heating treatment at 300°C. The obtained composite metal oxide film had a spherical structure with mass loading from 0.13 to 0.21 mg cm(-2). The structure
Catherine F Wise et al.
Dalton transactions (Cambridge, England : 2003), 44(32), 14265-14271 (2015-07-22)
We report a nickel complex containing a conjugated bis-dithiocarbazate ligand that is an active catalyst for the reduction of protons into hydrogen gas. Light-driven hydrogen generation is observed from a system containing this molecular nickel catalyst coupled with a fluorescein
Raziyeh Akbarzadeh et al.
Dalton transactions (Cambridge, England : 2003), 43(44), 16745-16753 (2014-10-04)
A NiS2 nanostructure with a protective layer of Ni(2+) and SO4(2-) ions around it has been successfully synthesized using the Ni(II)-Salen (Salen = N,N'-bis(salicylidene)ethylenediamine) complex via a simple solvothermal approach in the presence of anhydrous sodium thiosulfate (Na2S2O3) as sulfur

Articles

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service