377201
Manganese(II) oxide
powder, −60 mesh, 99%
Synonym(s):
Manganese monoxide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Assay
99%
form
powder
particle size
−60 mesh
density
5.45 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
O=[Mn]
InChI
1S/Mn.O
InChI key
VASIZKWUTCETSD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Manganese(II) oxide is an inorganic compound with the formula MnO that adopts the cubic rock-salt crystal structure with octahedral Mn2+ coordinating with octahedral O2-. Our 99% pure MnO is a green powder sieved through a 60 mesh (250 microns). Synthetically, it is a useful source of Mn2+ and is used in the battery industry as a precursor for manganese-containing cathode materials, like lithium nickel manganese cobalt oxide. It is also used in the semiconductor industry for fabricating thin-film transistors and other electronic devices. Because it is paramagneticat room temperature and considered to have good biocompatibility, MnO is alsoused in biomedical research in the development of contrast agents for magneticresonance imaging (MRI).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service