Skip to Content
Merck
All Photos(2)

Documents

376949

Sigma-Aldrich

Phosphorus(V) oxybromide

>95%

Synonym(s):

Phosphorus oxide bromide, Phosphoryl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
POBr3
CAS Number:
Molecular Weight:
286.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

>95%

form

solid

bp

192 °C (lit.)

mp

56 °C (lit.)

density

2.82 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

BrP(Br)(Br)=O

InChI

1S/Br3OP/c1-5(2,3)4

InChI key

UXCDUFKZSUBXGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of phosphorus pentabromide with phosphorus pentoxide at elevated temperatures affords phosphorus(V) oxybromide. Its Raman spectrum has been studied and the molecule has been reported to exhibit point group symmetry C3v. Phosphorus(V) oxybromide (Phosphorus oxide bromide) participates in the preparation of 1:1 addition ionic compound having melting point at 154·0°C, via reaction with gallium tribromide. This ionic compound contains non-metal cation POBr2+.

Application

Phosphorus(V) oxybromide may be used as brominating reagent in the synthesis of 2,4,6-tribromopyrimidine. It may be used in the preparation of bromoheterocyclic compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorus oxybromide as a brominating agent; bromopyrimidines.
D R V GOLDING et al.
The Journal of organic chemistry, 12(2), 293-293 (1947-03-01)
The possible existence of the cation POBr2+ and some properties of phosphorus oxybromide, gallium tribromide, and their ionic 1: 1 addition compound.
Greenwood NN and Worrall IJ.
J. Inorg. Nucl. Chem., 6(1), 34-41 (1958)
90. The heats of hydrolysis of some halides and oxyhalides of phosphorus.
Charnley T and Skinner HA.
Journal of the Chemical Society, 450-452 (1953)
A facile halogenation of some hydroxyheterocycles using triphenylphosphine and N-halosuccinimide.
Sugimoto O, et al.
Tetrahedron Letters, 40(42), 7477-7478 (1999)
The Raman spectrum and the molecular structure of phosphorus oxybromide.
Gerding H and van Driel M.
Rec. Trav. Chim., 61(6), 419-424 (1942)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service