Skip to Content
Merck
All Photos(2)

Key Documents

367079

Sigma-Aldrich

Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(OH)CH2CH2CO2(CH2)17CH3
CAS Number:
Molecular Weight:
530.86
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

mp

50-52 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C35H62O3/c1-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-26-38-32(36)25-24-29-27-30(34(2,3)4)33(37)31(28-29)35(5,6)7/h27-28,37H,8-26H2,1-7H3

InChI key

SSDSCDGVMJFTEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate is a propionate based phenolic antioxidant that can be used as a polymeric stabilizer by butylphenol functionalities.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discoloration of polymers by phenolic antioxidants.
Posp??sil J, et al.
Polymer Degradation and Stability, 77(3), 531-538 (2002)
E Niki et al.
The Journal of biological chemistry, 260(4), 2191-2196 (1985-02-25)
Inhibition of the oxidation of methyl linoleate and soybean phosphatidylcholine in homogeneous solution and in aqueous dispersion by four chain-breaking antioxidants, vitamin E (alpha-tocopherol), 2,2,5,7,8-pentamethyl-6-chromanol, 2,6-di-tert-butyl-4-methylphenol, and stearyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, was studied to examine the effect of the phytyl side chain
Determination of antioxidant migration levels from low-density polyethylene films into food simulants.
Dopico-Garc?a MS, et al.
Journal of Chromatography A, 1018(1), 53-62 (2003)
I Cooper et al.
Food additives and contaminants, 15(1), 72-78 (1998-04-16)
Many additives used in plastics materials and articles intended for food contact are expected to be assigned specific migration limits (SMLs) in a future amendment to EC Directive 90/128/EEC. In order to demonstrate compliance with these restrictions, specific migration tests
Effect of secondary structure on physical behaviour and performance of hindered phenolic antioxidants in polypropylene.
Tochacek J
Polymer Degradation and Stability, 86(2), 385-389 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service