Skip to Content
Merck
All Photos(2)

Documents

365319

Sigma-Aldrich

Zinc-Copper couple

powder

Synonym(s):

Zinc, compd. with copper

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

419-420 °C (lit.)

SMILES string

[Cu].[Zn]

InChI

1S/Cu.Zn

InChI key

TVZPLCNGKSPOJA-UHFFFAOYSA-N

Application


  • Unravelling rechargeable zinc-copper batteries by a chloride shuttle in a biphasic electrolyte: This article explores the use of zinc-copper redox couples in rechargeable batteries, focusing on their high reversible capacity and efficiency. (Xu et al., 2023).

  • Rapid and Safe Continuous-Flow Simmons-Smith Cyclopropanation using a Zn/Cu Couple Column: This research presents a continuous-flow Simmons-Smith cyclopropanation method utilizing a zinc-copper couple column reactor for safer and faster reactions. (Nova-Fernández et al., 2024).

  • Efficient removal of aniline by micro-scale zinc-copper (mZn/Cu) bimetallic particles in acidic solution: The study demonstrates the effective degradation of aniline using micro-scale zinc-copper bimetallic particles, focusing on the oxidation degradation mechanism. (Li et al., 2019).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service