Skip to Content
Merck
All Photos(3)

Key Documents

364258

Sigma-Aldrich

3-Aminopropyl-functionalized silica gel

40-63 μm, extent of labeling: ~1 mmol/g NH2 loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

form

powder

extent of labeling

~1 mmol/g NH2 loading
~9% functionalized

surface area

550 m2/g

matrix

irregular silica particle platform

matrix active group

NH2 phase

particle size

40-63 μm

pore size

60 Å pore size

SMILES string

O[SiH3].CCCN

InChI key

ZPPBNRTUMSCLLZ-UHFFFAOYSA-N

General description

The product is silica gel functionalized with amino groups. The product (3-Aminopropyl-functionalized silica gel) is 9% functionalized and has 1mmol/g of NH2 loading (extent of labelling). It participates as catalyst in the Knoevenagel condensation reactions.

Application

3-Aminopropyl-functionalized silica gel may be used in the following studies:
  • For the separation of polychlorinated biphenyls (PCBs) from lipids by combined supercritical fluid extraction and separation.
  • Preparation of the tin complex immobilized on silica gel, a novel, green, highly efficient and heterogeneous reusable catalyst.
  • Preparation of silica gel bearing isonicotinamide groups.
  • Synthesis of a novel catalytic system including Pd(OAc)2 attached to a bis(oxazoline) ligand that is covalently bonded to 3-mercaptopropyl-functionalized silica gel.
  • Useful as a heterogeneous, reusable catalyst for the Knoevenagel condensation.
  • For low- and medium-pressure reversed-phase liquid chromatography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ramin Ghahremanzadeh et al.
Dalton transactions (Cambridge, England : 2003), 43(42), 15791-15797 (2014-09-13)
In the present study, a tin complex immobilized on silica gel as a novel, green, highly efficient and heterogeneous reusable catalyst was synthesized by grafting 2-amino benzamide onto the silica gel surface as a result of the reaction between isatoic
E G Alley et al.
Journal of AOAC International, 78(4), 1051-1054 (1995-07-01)
We present a method that combines the extraction of polychlorinated biphenyls (PCBs) and their separation from relatively large quantities of fat in biological matrixes by combined supercritical fluid extraction and separation. Cyano-functionalized silica gel, silica gel, aluminum oxide, Florisil, 3-amino-propyl-functionalized
H Gruber-Woelfler et al.
Journal of catalysis, 286(9), 30-40 (2012-08-10)
The synthesis and characterization of a novel catalytic system including Pd(OAc)(2) attached to a bis(oxazoline) (=BOX) ligand that is covalently bonded to 3-mercaptopropyl-functionalized silica gel is presented. The catalyst was tested for Suzuki-Miyaura reactions of different aryl halides with phenylboronic
Journal of the Chemical Society. Perkin Transactions 1, 105-105 (1989)
Amino groups immobilized on silica gel: an efficient and reusable heterogeneous catalyst for the Knoevenagel condensation.
Angeletti E, et al.
Journal of the Chemical Society. Perkin Transactions 1, 1, 105-107 (1989)

Articles

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service