Skip to Content
Merck
All Photos(1)

Documents

359661

Sigma-Aldrich

H-Lys(Boc)-OH

≥95%, for peptide synthesis

Synonym(s):

Nε-Boc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONH(CH2)4CH(NH2)COOH
CAS Number:
Molecular Weight:
246.30
Beilstein:
2417626
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

H-Lys(Boc)-OH, ≥95%

Quality Level

Assay

≥95%

form

powder

optical activity

[α]20/D +18°, c = 1 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

250 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCC[C@H](N)C(O)=O

InChI

1S/C11H22N2O4/c1-11(2,3)17-10(16)13-7-5-4-6-8(12)9(14)15/h8H,4-7,12H2,1-3H3,(H,13,16)(H,14,15)/t8-/m0/s1

InChI key

VVQIIIAZJXTLRE-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

H-Lys(Boc)-OH also known as Nε-Boc-L-lysine, is commonly used in solution phase peptide synthesis.

Application

H-Lys(Boc)-OH can be used to prepare pentafluorophenyl esters which further used to synthesize β-peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution phase synthesis of beta-peptides using micro reactors
P Watts
Tetrahedron, 58, 5427-5439 (2002)
Maria Moccia et al.
International journal of pharmaceutics, 397(1-2), 179-183 (2010-07-06)
A novel nucleic acid compaction device based on a positively-charged alpha,epsilon-poly-l-lysine was realized for the first time. The polycationic peptide was obtained by assembling Fmoc and Boc orthogonally protected l-lysine monomers by solid phase synthesis. The route to the novel
April Case et al.
Analytical biochemistry, 338(2), 237-244 (2005-03-05)
Tissue transglutaminase (TGase) is a Ca(2+)-dependent enzyme that catalyzes cross-linking of intracellular proteins through a mechanism that involves isopeptide bond formation between Gln and Lys residues. In addition to its transamidation activity, TGase can bind guanosine 5'-triphosphate (GTP) and does
Ognyan K Argirov et al.
Biochimica et biophysica acta, 1620(1-3), 235-244 (2003-02-22)
Proteins are subject of posttranslational modification by sugars and their degradation products in vivo. The process is often referred as glycation. L-Dehydroascorbic acid (DHA), an oxidation product of L-ascorbic acid (vitamin C), is known as a potent glycation agent. A
J T Sparrow et al.
Peptide research, 9(6), 297-304 (1996-11-01)
We have synthesized a hydrophilic crosslinked aminoalkyl polydimethylacrylamide-beaded support upon which peptides have been assembled using standard Fmoc chemistry in automated batch-wise equipment. The resin was prepared by the free radical-initiated co-polymerization of N,N-dimethylacryl-amide, N,N'-bisacrylyl-1,3-diaminopropane and a functional monomer N-methacrylyl-1,3-diaminopropane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service