Skip to Content
Merck
All Photos(1)

Documents

348287

Sigma-Aldrich

(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution

60 wt. % in H2O

Synonym(s):

3-chloro-2-hydroxy-propyltrimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OH)CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
188.10
Beilstein:
6576172
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

60 wt. % in H2O

refractive index

n20/D 1.4541

density

1.154 g/mL at 25 °C

SMILES string

[Cl-].C[N+](C)(C)CC(O)CCl

InChI

1S/C6H15ClNO.ClH/c1-8(2,3)5-6(9)4-7;/h6,9H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

CSPHGSFZFWKVDL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution (CHTAC) can be used:
  • As a cation-generating agent for cellulose cationization by exhaustion method.
  • To resolve 2,2′-dihydroxy-1,1′-binaphthyl enantiomers.
  • To synthesize cationic glycogen (Cat Gly).
  • As a quaternizing agent for quaternization of N-aryl chitosan derivatives.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepak Sharma et al.
Carbohydrate polymers, 232, 115731-115731 (2020-01-19)
Galactomannans are branched natural biopolymers possessing good biocompatibility, biodegradability and sustainability leading to diverse industrial applications. In the present study, quaternization of galactomannan derived from Cassia tora gum was investigated by adopting Taguchi's approach of robust design of experiments using
Quaternization of N-aryl chitosan derivatives: synthesis, characterization, and antibacterial activity
Sajomsang W, et al.
Carbohydrate Research, 344(18), 2502-2511 (2009)
Effect of cationization on adsorption of silver nanoparticles on cotton surfaces and its antibacterial activity
Khalil-Abad M, et al.
Cellulose, 16(6), 1147-1147 (2009)
Reaction efficiency for cellulose cationization using 3-chloro-2-hydroxypropyl trimethyl ammonium chloride
Hashem M, et al.
Textile Res., 73(11), 1017-1023 (2003)
Synthesis, characterization and flocculation characteristics of cationic glycogen: a novel polymeric flocculant
Pal S, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 289(1-3), 193-199 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service