Skip to Content
Merck
All Photos(2)

Key Documents

348155

Sigma-Aldrich

Methyl 4-(bromomethyl)benzoate

98%

Synonym(s):

α-Bromo-p-toluic acid methyl ester, 4-(Bromomethyl)benzoic acid methyl ester, 4-(Carbomethoxy)benzyl bromide, 4-(Methoxycarbonyl)benzyl bromide, Methyl α-bromo-p-toluate, Methyl p-(bromomethyl)benzoate, p-(Bromomethyl)benzoic acid methyl ester, p-(Carbomethoxy)benzyl bromide, p-(Methoxycarbonyl)benzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4CO2CH3
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

130-135 °C/2 mmHg (lit.)

mp

57-58 °C (lit.)

SMILES string

COC(=O)c1ccc(CBr)cc1

InChI

1S/C9H9BrO2/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-5H,6H2,1H3

InChI key

NLWBJPPMPLPZIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 4-(bromomethyl)benzoate is an ester. It is a lachrymator and an important drug intermediate. Ester group is slightly twisted out of the plane of the central aromatic ring in methyl 4-(bromomethyl)benzoate.

Application

Methyl 4-(bromomethyl)benzoate may be used in the synthesis of 9-O-(4-carboxybenzyl)berberine (CBB).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adel A Marzouk et al.
Bioorganic chemistry, 101, 104020-104020 (2020-07-01)
New imidazolidindiones and tetra-substituted imidazole derivatives were designed, synthesized, and evaluated for the anticonvulsant activity through pentylenetetrazole (PTZ)-induced seizures and maximal electroshock (MES) tests using valproate sodium and phenytoin sodium as reference drugs, respectively. Most of the target compounds showed
Methyl 4-(bromomethyl) benzoate.
Yathirajan HS, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(3), o1334-o1335 (2007)
Jin-Xiang Chen et al.
Bioorganic & medicinal chemistry letters, 22(23), 7056-7059 (2012-10-23)
9-O-(4-carboxybenzyl)berberine (CBB) 1 was synthesized from the reaction of berberrubine with methyl 4-(bromomethyl)benzoate and subsequent hydrolysis. Its Cu(II) complex 2 was prepared from the reaction with Cu(NO(3))(2)·3H(2)O, and established as [Cu(CBB)(2)](NO(3))(2)·2H(2)O by means of (1)H NMR, UV, IR, elemental analysis
Jia-Rong Liu et al.
Biochemical pharmacology, 163, 458-471 (2019-03-20)
Glioblastoma is the most fatal type of primary brain cancer, and current treatments for glioblastoma are insufficient. HDAC6 is overexpressed in glioblastoma, and siRNA-mediated knockdown of HDAC6 inhibits glioma cell proliferation. Herein, we report a high-selective HDAC6 inhibitor, J22352, which
Kai Yao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1080, 50-58 (2018-02-25)
An immunoaffinity clean-up material based on a monoclonal antibody (mAb) has been prepared for concentrating and purifying bisphenol A (BPA) in 14 kinds of foodstuffs at trace level. Haptens and immunogen of bisphenol A have been synthesized and comprehensively characterized.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service