Skip to Content
Merck
All Photos(3)

Key Documents

344427

Sigma-Aldrich

Iron(II) D-gluconate dihydrate

98%

Synonym(s):

D-Gluconic acid iron(II) salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOCH2[CH(OH)]4CO2]2Fe · 2H2O
CAS Number:
Molecular Weight:
482.17
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

optical activity

[α]25/D +6.7°, c = 1 in H2O

mp

188 °C (dec.) (lit.)

SMILES string

O.O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Fe]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Fe.2H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;;/h2*2-5,7-11H,1H2,(H,12,13);;2*1H2/q;;+2;;/p-2/t2*2-,3-,4+,5-;;;/m11.../s1

InChI key

OKGNXSFAYMSVNN-SYAJEJNSSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey E Nowak et al.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies, 13(5), e323-e329 (2012-07-05)
We previously demonstrated that altered zinc homeostasis is an important feature of pediatric sepsis, thus raising the possibility of zinc supplementation as a therapeutic strategy in sepsis. Herein, we tested the hypothesis that prophylactic zinc supplementation would be beneficial in
Michael T Rose et al.
Planta, 236(4), 959-973 (2012-04-25)
Plants are routinely subjected to multiple environmental stresses that constrain growth. Zinc (Zn) deficiency and high bicarbonate are two examples that co-occur in many soils used for rice production. Here, the utility of metabolomics in diagnosing the effect of each
Volker Behrends et al.
The Journal of biological chemistry, 288(21), 15098-15109 (2013-04-11)
Metabolic footprinting of supernatants has been proposed as a tool for assigning gene function. We used NMR spectroscopy to measure the exometabolome of 86 single-gene transposon insertion mutant strains (mutants from central carbon metabolism and regulatory mutants) of the opportunistic
[Item 232--Facial dermatoses: acne].
Annales de dermatologie et de venereologie, 139(11 Suppl), A192-A196 (2012-12-05)
Angel A Yanagihara et al.
PloS one, 7(12), e51368-e51368 (2012-12-20)
Chironex fleckeri (Australian box jellyfish) stings can cause acute cardiovascular collapse and death. We developed methods to recover venom with high specific activity, and evaluated the effects of both total venom and constituent porins at doses equivalent to lethal envenomation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service