Skip to Content
Merck
All Photos(3)

Key Documents

339792

Sigma-Aldrich

3,4-Dibutoxy-3-cyclobutene-1,2-dione

98%

Synonym(s):

Dibutyl squarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3O]2C4(=O)2
CAS Number:
Molecular Weight:
226.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

139 °C/0.5 mmHg (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

ether
ketone

SMILES string

CCCCOC1=C(OCCCC)C(=O)C1=O

InChI

1S/C12H18O4/c1-3-5-7-15-11-9(13)10(14)12(11)16-8-6-4-2/h3-8H2,1-2H3

InChI key

XBRWELTXMQSEIN-UHFFFAOYSA-N

General description

3,4-Dibutoxy-3-cyclobutene-1,2-dione is the dialkyl ester of squaric acid.

Application

3,4-Dibutoxy-3-cyclobutene-1,2-dione was used in the synthesis of novel semisquarylium dye, useful for highly selective Hg2+ sensing in aqueous media. It was also employed in the synthesis of 3-butoxy-4-(1,3,3-trimethyl-2,3-dihydro-1H-2-indolylidenemethyl)-3-cyclobutene-1,2-dione, intermediate for the synthesis of squaryl dyes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular and crystal structure of 3-butoxy-4-(1, 3, 3-trimethyl-2, 3-dihydro-1H-2-indolylidenemethyl)-3-cyclobutene-1, 2-dione and its thio analog.
Shishkina SV, et al.
Journal of Structural Chemistry, 46(1), 154-158 (2005)
Amy J McMichael et al.
Dermatology nursing, 16(4), 333-336 (2004-10-09)
Though many therapies exist for alopecia areata, one of the most unique is topical sensitization. By altering the pathogenic inflammatory response with few side effects, sensitizers offer an attractive treatment option for many patients with alopecia areata, including those who
Eileen Tan et al.
International journal of dermatology, 41(11), 748-753 (2002-11-28)
Alopecia areata is believed to be an autoimmune condition with a worldwide occurrence. It usually presents as patchy, nonscarring hair loss. There is a paucity of clinical data in Asians. To study the epidemiology, clinical aspects, associations, and treatment of
C Ajith et al.
Journal of drugs in dermatology : JDD, 5(3), 262-266 (2006-04-01)
Immunotherapy with sqauric acid dibutyl ester (SADBE) is a well-accepted therapy for alopecia areata. To study efficacy, safety, and factors influencing the outcome in the treatment of alopecia areata. During a 4-year period, 70 patients of alopecia areata, unresponsive to
F Dall'oglio et al.
The Journal of dermatological treatment, 16(1), 10-14 (2005-05-18)
Severe alopecia areata (AA) may have a chronic relapsing course and is often resistant to current treatments. The aim of our study was to evaluate whether topical immunotherapy with squaric acid dibutylester (SADBE) is able to improve the course of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service