Skip to Content
Merck
All Photos(1)

Documents

332143

Sigma-Aldrich

L-Glutamic acid-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)2CH(15NH)CO2H
CAS Number:
Molecular Weight:
148.12
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

Assay

99% (CP)

form

powder

optical activity

[α]25/D +31.0°, c = 2 in 5 M HCl

technique(s)

bio NMR: suitable

mp

205 °C (dec.) (lit.)

mass shift

M+1

SMILES string

[15NH2]C(CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i6+1

InChI key

WHUUTDBJXJRKMK-OGWWSMAPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kishore K Mahalingan et al.
Nature structural & molecular biology, 27(9), 802-813 (2020-08-05)
Glutamylation, introduced by tubulin tyrosine ligase-like (TTLL) enzymes, is the most abundant modification of brain tubulin. Essential effector proteins read the tubulin glutamylation pattern, and its misregulation causes neurodegeneration. TTLL glutamylases post-translationally add glutamates to internal glutamates in tubulin carboxy-terminal
E Driehuis et al.
PloS one, 15(5), e0231588-e0231588 (2020-05-19)
We have recently established a protocol to grow wildtype human oral mucosa organoids. These three-dimensional structures can be maintained in culture long-term, do not require immortalization, and recapitulate the multilayered composition of the epithelial lining of the oral mucosa. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service