Skip to Content
Merck
All Photos(2)

Documents

325678

Sigma-Aldrich

Gadolinium(III) acetate hydrate

99.9% trace metals basis

Synonym(s):

Gadolinium acetate hydrate, Gadolinium triacetate hydrate, Gadolinium(3+) tris(acetate) hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
334.38 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

crystalline powder

reaction suitability

core: gadolinium
reagent type: catalyst

SMILES string

O.CC(=O)O[Gd](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Gd.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

GXCUBNRUECGTSC-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sugata Barui et al.
Nanomaterials (Basel, Switzerland), 10(6) (2020-06-18)
Zinc oxide nanocrystals (ZnO-NCs) doped with transition metal elements or rare earth elements can be probed for magnetic resonance imaging to be used as a molecular imaging technique for accurate diagnosis of various diseases. Herein, we use Mn as a
Yuichi Kitagawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(1), 264-269 (2020-07-04)
A design for an effective molecular luminescent thermometer based on long-range electronic coupling in lanthanide coordination polymers is proposed. The coordination polymers are composed of lanthanide ions EuIII and GdIII , three anionic ligands (hexafluoroacetylacetonate), and a chrysene-based phosphine oxide
Haruki Minami et al.
Scientific reports, 10(1), 18917-18917 (2020-11-05)
DNA-based materials have attracted much attention due to their unique photo-functional properties and potential applications in various fields such as luminescent and biological systems, nanodevices, etc. In this study, the photophysical properties of a chiral Eu(III) complex, namely (Eu(D-facam)3), within
Krishnaiah Damarla et al.
Langmuir : the ACS journal of surfaces and colloids, 34(34), 10081-10091 (2018-07-29)
Nanoemulsions (NEs) comprising ionic liquids (ILs); ethanolammonium formate (HO-EOAF), proliniumisopropylester dioctylsulfosuccinate ([ProC3][AOT]), and 1-butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, ([Bmim][NTf2]) as insoluble hydrophilic, surface active, and hydrophobic components have been constructed. This novel class of colloidal formulations exhibited several contrasting properties vis-à-vis conventional water-in-oil
Shuai Zha et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(5), e2002919-e2002919 (2021-03-16)
Using Epstein-Barr virus (EBV)-induced cancer cells and HeLa cells as a comparative study model, a novel and safe dual-EBV-oncoproteins-targeting pH-responsive peptide engineering, coating, and guiding approach to achieve precision targeting and treatment strategy against EBV-associated cancers is introduced. Individual functional

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service