Skip to Content
Merck
All Photos(1)

Key Documents

324248

Sigma-Aldrich

4-Bromo-1,8-naphthalic anhydride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5BrO3
CAS Number:
Molecular Weight:
277.07
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

mp

217-219 °C (lit.)

SMILES string

Brc1ccc2C(=O)OC(=O)c3cccc1c23

InChI

1S/C12H5BrO3/c13-9-5-4-8-10-6(9)2-1-3-7(10)11(14)16-12(8)15/h1-5H

InChI key

DTUOTSLAFJCQHN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youlu Chu et al.
International journal of biological macromolecules, 153, 46-54 (2020-03-01)
Fluorescent nanoprobe with good water dispersibility was synthesized by the coupling of fluorescent 1,8-naphthalimide dye (NANI) as well as biocompatible poly (ethylene glycol) (PEG) to cellulose nanocrystals (CNC). FTIR, TGA and XPS analysis confirmed the successful covalent conjugation of NANI
Piyali Jana et al.
International journal of biological macromolecules, 161, 1149-1160 (2020-06-20)
The study emphasized on the development of an efficient, receptor-targeted non-viral gene delivery vehicle for gene therapy of triple negative breast cancer (TNBC). Here, naturally abundant guar gum based non-viral carrier was developed through conjugating by low molecular weight polyethylenimine
Hao Liu et al.
Macromolecular rapid communications, 41(13), e2000123-e2000123 (2020-05-14)
Substituted naphthalimide (NI) moieties are highly versatile and newly recognized aggregation-induced emission (AIE) building blocks for many potentially useful smart molecules, polymers, and nanoparticles. However, the introduction of NI fluorophore into cross-linked polymeric networks to prepare AIE-active hydrogels still remains
Fang Dai et al.
Free radical research, 52(11-12), 1288-1295 (2018-08-22)
Hydrogen peroxide (H2O2) produced from mitochondria has attracted much attention on account of its physiological and pathological functions. Therefore, monitoring mitochondrial H2O2 levels in living cells is of great significance for understanding its functions. We report here a mitochondria-targeted H2O2
Xinhua Cao et al.
Physical chemistry chemical physics : PCCP, 17(48), 32297-32303 (2015-11-20)
A two-component light-harvesting organogel containing a naphthalimide-based gelator (1) as a donor and a phosphorescent Ir(III) complex [Ir(bt)2(acac)] (bt = 2-phenylbenzothiazole and acac = acetylacetone) (Ir) as an acceptor was used to produce white-light-emitting organogels. The addition of complex Ir

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service