Skip to Content
Merck
All Photos(1)

Key Documents

317675

Sigma-Aldrich

Ethyl oleate

technical grade, 70%

Synonym(s):

Oleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOC2H5
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727318
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical grade

Assay

70%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

205-208 °C/760 mmHg
216-218 °C/15 mmHg

mp

−32 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-

InChI key

LVGKNOAMLMIIKO-QXMHVHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Khushwinder Kaur
Journal of the science of food and agriculture, 95(13), 2718-2728 (2014-11-25)
Inclusion complex formation between β-cyclodextrin (β-CD) and suitable guest molecules such as cholesterol (Ch) has regularly been exploited to design self-assembled structures. In the present study an effective nanoemulsion medium (lecithin/Tween 80/ethyl oleate/water) was selected for solubilizing and stabilizing Ch
Michael D Hardy et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8187-8192 (2015-06-24)
Cell membranes are dynamic structures found in all living organisms. There have been numerous constructs that model phospholipid membranes. However, unlike natural membranes, these biomimetic systems cannot sustain growth owing to an inability to replenish phospholipid-synthesizing catalysts. Here we report
H H Orak et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 18(4), 391-402 (2012-04-24)
Antioxidant activity, colour and some nutritional properties of hot air and freeze-dried strawberry tree (Arbutus unedo L.) fruits were investigated. Additionally, the effects of two pre-treatments, namely ethyl oleate and water blanching, were compared in terms of drying characteristics. For
Andreas Braun et al.
Microbial cell factories, 11, 106-106 (2012-08-11)
Yarrowia lipolytica efficiently metabolizes and assimilates hydrophobic compounds such as n-alkanes and fatty acids. Efficient substrate uptake is enabled by naturally secreted emulsifiers and a modified cell surface hydrophobicity and protrusions formed by this yeast. We were examining the potential
Thomas S Muenz et al.
Die Naturwissenschaften, 99(5), 421-425 (2012-03-20)
Social work force distribution in honeybee colonies critically depends on subtle adjustments of an age-related polyethism. Pheromones play a crucial role in adjusting physiological and behavioral maturation of nurse bees to foragers. In addition to primer effects of brood pheromone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service