Skip to Content
Merck
All Photos(1)

Key Documents

317640

Sigma-Aldrich

mono-Methyl phthalate

97%

Synonym(s):

Monomethyl phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO2C)C6H4CO2CH3
CAS Number:
Molecular Weight:
180.16
Beilstein:
2048766
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

82-84 °C (lit.)

SMILES string

COC(=O)c1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-13-9(12)7-5-3-2-4-6(7)8(10)11/h2-5H,1H3,(H,10,11)

InChI key

FNJSWIPFHMKRAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Xin Wang et al.
Environmental research, 142, 1-9 (2015-06-19)
Exposure to phthalates has been demonstrated to have adverse effects on male reproduction in animal studies, but findings in human studies have been inconsistent. We recruited 1040 men from the Reproductive Center of Tongji Hospital in Wuhan, China from March
Da-Wei Liang et al.
Applied microbiology and biotechnology, 74(1), 221-229 (2006-11-11)
Results of batch experiments on the denitrifying degradation of dimethyl phthalate (DMP) was most favorable at pH 7-9 and 30-35 degrees C. DMP was first degraded to monomethyl phthalate (MMP), which was in turn degraded to phthalate before complete mineralization.
Manori J Silva et al.
Archives of toxicology, 79(11), 647-652 (2005-07-05)
In assessment of exposure to environmental contaminants, the use of unconventional matrices is becoming an increasingly important area of research. Saliva is one of the most promising alternative matrices because its collection is easy, noninvasive, and inexpensive. In this study
Jia Bao et al.
Archives of environmental contamination and toxicology, 69(2), 202-207 (2015-03-10)
In 2011, Taiwan authorities reported that two phthalates, including di-(2-ethylhexyl) phthalate and di-iso-nonyl phthalate, were intentionally introduced into a variety of foods and beverages during the course of 15 years. However, little is known about body burdens of phthalate contaminations in
Saou-Hsing Liou et al.
Journal of hazardous materials, 277, 169-179 (2014-03-19)
This 5-month study contains two parts: (1) to monitor the concentrations of 11 phthalate esters metabolites (PAEMs) and two beta-agonists in human urine samples collected from a small group of consented participants including 16 females and five males; and (2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service