Skip to Content
Merck
All Photos(1)

Key Documents

308188

Sigma-Aldrich

Benzoyl isocyanate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONCO
CAS Number:
Molecular Weight:
147.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

refractive index

n20/D 1.5514 (lit.)

bp

94-96 °C/21 mmHg (lit.)

density

1.171 g/mL at 25 °C (lit.)

functional group

amine
isocyanate
phenyl

storage temp.

−20°C

SMILES string

O=C=NC(=O)c1ccccc1

InChI

1S/C8H5NO2/c10-6-9-8(11)7-4-2-1-3-5-7/h1-5H

InChI key

LURYMYITPCOQAU-UHFFFAOYSA-N

General description

The reaction mechanism of benzoyl isocyanate with 6-benzyl-6-azabicyclo[2.2.1]hept-2-ene was studied.

Application

Benzoyl isocyanate was used as a capping reagent for low-reactivity hydroxy groups in the solid-phase-supported oligosaccharide synthesis.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangyang Wu et al.
The Journal of organic chemistry, 69(6), 1853-1857 (2004-04-03)
Solid-phase-supported oligosaccharide synthesis of a core N-glycan tetrasaccharide and of a trisaccharide containing the Galili antigen is reported. The synthesis is based on a hydroxymethylbenzyl benzoate spacer-linker system attached to the Merrifield resin, O-Fmoc-protected O-glycosyl trichloroacetimidates as glycosyl donors, and
P Madhusudhana Reddy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 186, 8-16 (2017-06-11)
Sensing of F
Cong Zhang et al.
The journal of physical chemistry. A, 114(8), 2913-2919 (2010-02-09)
Reaction mechanisms of the 6-benzyl-6-azabicyclo[2.2.1]hept-2-ene with benzoyl isocyanate have been investigated using density functional theory (DFT) at the B3LYP/6-31G(d,p) level of theory. The reaction proceeding along six competitive channels includes two categories. That is, two channels are formally [3,3]-sigmatropic rearrangements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service