All Photos(1)
About This Item
Linear Formula:
[C2H5CH(CH3)]2NH
CAS Number:
Molecular Weight:
129.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.41 (lit.)
bp
135 °C/765 mmHg (lit.)
density
0.753 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCC(C)NC(C)CC
InChI
1S/C8H19N/c1-5-7(3)9-8(4)6-2/h7-9H,5-6H2,1-4H3
InChI key
OBYVIBDTOCAXSN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Di-sec-butylamine undergoes exothermic exchange reaction with the cluster [N,N,N′,N′-tetramethyl bis-endo-BCH-2,5-diamide...H+...di-isopropylamine].
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
73.8 °F - closed cup - (own results)
Flash Point(C)
23.2 °C - closed cup - (own results)
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Matthias Witt et al.
Physical chemistry chemical physics : PCCP, 7(5), 1065-1072 (2005-03-07)
The ligand exchange reactions of the proton-bound trimer [(DMF)2...H+...H2NCH3] of dimethylformamide (DMF) and methylamine and of proton-bound dimers [diamide...H+...amine] of three aliphatic primary diamides, succinic diamide, adipinic diamide, and maleic diamide, and of three tertiary diamides N,N,N',N'-tetramethyl succinic diamide, N,N,N',N'-tetramethyl
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service