Skip to Content
Merck
All Photos(1)

Key Documents

306622

Sigma-Aldrich

4-Chloro-3-fluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)OH
CAS Number:
Molecular Weight:
146.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%

Quality Level

Assay

98%

form

solid

bp

84 °C/44 mmHg (lit.)

mp

54-56 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear, colorless

functional group

chloro
fluoro

SMILES string

Oc1ccc(Cl)c(F)c1

InChI

1S/C6H4ClFO/c7-5-2-1-4(9)3-6(5)8/h1-3,9H

InChI key

XLHYAEBESNFTCA-UHFFFAOYSA-N

General description

4-Chloro-3-fluorophenol is hydroxylated at both ortho positions to yield different products.

Application

4-Chloro-3-fluorophenol was used in the synthesis of 4-chloro-3-fluoro catechol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ridder et al.
European journal of biochemistry, 257(1), 92-100 (1998-11-03)
The influence of various C4/C5 substituents in catechol (1,2-dihydroxybenzene) derivatives on the overall rate of conversion by catechol-1,2-dioxygenase from Pseudomonas putida (arvilla) C1 was investigated. Using catechol, 4-methylcatechol, 4-fluorocatechol, 4-chlorocatechol, 4-bromocatechol, 4,5-difluorocatechol and 4-chloro-5-fluorocatechol, it could be demonstrated that substituents
S Peelen et al.
European journal of biochemistry, 227(1-2), 284-291 (1995-01-15)
This study describes the regioselective hydroxylation and the rates of conversion of a series of fluorinated phenol derivatives by phenol hydroxylase from the yeast Trichosporon cutaneum. The natural logarithm of the kcat value for the conversion of the phenolic substrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service