Skip to Content
Merck
All Photos(1)

Documents

303011

Sigma-Aldrich

Heptane-d16

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)5CD3
CAS Number:
Molecular Weight:
116.30
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.5 (vs air)

Quality Level

100
200

vapor pressure

40 mmHg ( 20 °C)

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

autoignition temp.

595 °F

technique(s)

NMR: suitable

refractive index

n20/D 1.3844 (lit.)

bp

98 °C (lit.)

mp

-91 °C (lit.)

density

0.794 g/mL at 25 °C (lit.)

mass shift

M+16

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2

InChI key

IMNFDUFMRHMDMM-NEBSKJCTSA-N

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution phase coordination chemistry of trivalent lanthanide and actinide cations with bis (2, 4, 4-trimethylpentyl) dithiophosphinic acid.
Jensen MP, et al.
Journal of Nuclear Science and Technology (Abingdon, United Kingdom), 39, 255-258 (2002)
The polarizability of some deuterated hydrocarbons.
LeGrand DG and Gaines Jr GL.
The Journal of Physical Chemistry, 98(18), 4842-4844 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service