Skip to Content
Merck
All Photos(1)

Key Documents

299405

Sigma-Aldrich

Magnesium

rod, diam. 6 mm, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 621 °C)

Quality Level

Assay

≥99.9% trace metals basis

form

rod

autoignition temp.

950 °F

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

diam.

6 mm

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium is the most common mineral found in the human body. It is a cofactor for more than 300 enzymes and is known to regulate physiological functions like blood pressure, muscle contraction, and neuromuscular conduction. It can be used in the preparation of many biomaterials for biomedical applications.

Application

Magnesium can be used:
  • As a working electrode in the study of the electrochemical behavior of Li+ ions.
  • As an electrode material in the electropolymerization of di-n-hexyldichlorosilane.

Quantity

10 g = 2 × 100 mm; 50 g = 10 × 100 mm

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The importance of magnesium in clinical healthcare
Schwalfenberg GK and Genuis SJ
Scientifica, 2017 (2017)
Electrodeposition of Li and electrochemical formation of Mg--Li alloys from the eutectic LiCl-KCl
Chen Z, et al.
Journal of alloys and compounds, 464(1-2), 174-178 (2008)
Factors influencing the electroreductive polymerization of di-n-hexyldichlorosilane
Martins L, et al.
Applied Organometallic Chemistry, 16(2), 76-83 (2002)
Magnesium and human health: perspectives and research directions
Al Alawi AM, et al.
International Journal of Endocrinology, 2018 (2018)
Review of magnesium-based biomaterials and their applications
Sezer N, et al.
Journal of Magnesium and Alloys, 6(1), 23-43 (2018)

Articles

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service