Skip to Content
Merck
All Photos(1)

Key Documents

295450

Sigma-Aldrich

2-Methylpropane

99%

Synonym(s):

Isobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CH
CAS Number:
Molecular Weight:
58.12
Beilstein:
1730720
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.01 (21 °C, vs air)

Quality Level

vapor pressure

72.2 psi ( 37.7 °C)

Assay

99%

autoignition temp.

860 °F

expl. lim.

8.3 %

bp

−12 °C (lit.)

mp

−160 °C (lit.)

density

2.064 g/mL at 25 °C (lit.)

SMILES string

CC(C)C

InChI

1S/C4H10/c1-4(2)3/h4H,1-3H3

InChI key

NNPPMTNAJDCUHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Methylpropane can be used as a starting material to prepare:
  • Isobutylene by catalytic dehydrogenation using various catalysts.[1]
  • Isopropyl methyl ketone by formylation reaction with carbon monoxide in the presence of super acid catalyst.[2]
  • tert-Butylhydroperoxide (TBHP) by oxidation.[3]

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-117.4 °F - closed cup

Flash Point(C)

-83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The production and use of butadiene and isobutylene
Petroleum Chemistry (Neftekhimiya), 112(14), 84-84 (2015)
Effect of catalyst prereduction on the dehydrogenation of isobutane over chromia/alumina
Airaksinen S MK and Krause A OI
Industrial & Engineering Chemistry Research, 44(11), 3862-3868 (2005)
Superacid-Catalyzed Selective Formylation-Rearrangement of Isoalkanes with Carbon Monoxide to Branched Ketones
Olah GA, et al.
Angewandte Chemie (International ed. in English), 112(14), 2647-2648 (2000)
Laure Pison et al.
Physical chemistry chemical physics : PCCP, 17(45), 30328-30342 (2015-10-29)
The solubility of n-butane and 2-methylpropane (isobutane) in three ionic liquids - 1-(2-methylpropyl)-3-methylimidazolium bis(trifluoromethylsulfonyl)imide [(2mC3)C1im][Ntf2], 1-(3-methylbutyl)-3-methylimidazolium bis(trifluoromethylsulfonyl)imide [(3mC4)C1im][Ntf2] and 1-methyl-3-pentylimidazolium bis(trifluoromethylsulfonyl)imide [C5C1im][Ntf2] - has been measured at atmospheric pressure from 303 to 343 K. Isobutane is less soluble than n-butane
Vera Lepperhof et al.
PloS one, 9(9), e107363-e107363 (2014-09-17)
Cell loss after transplantation is a major limitation for cell replacement approaches in regenerative medicine. To assess the survival kinetics of induced pluripotent stem cell (iPSC)-derived cardiomyocytes (CM) we generated transgenic murine iPSC lines which, in addition to CM-specific expression

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service